Skip to Content
Merck
All Photos(2)

Documents

HPA006988

Sigma-Aldrich

Anti-ZMPSTE24 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CAAX prenyl protease 1 homolog antibody produced in rabbit, Anti-FACE-1 antibody produced in rabbit, Anti-Farnesylated proteins-converting enzyme 1 antibody produced in rabbit, Anti-Prenyl protein-specific endoprotease 1 antibody produced in rabbit, Anti-Zinc metalloproteinase Ste24 homolog antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

KTTTHVPPELGQIMDSETFEKSRLYQLDKSTFS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Immunogen

CAAX prenyl protease 1 homolog recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

ZMPSTE24 (Zinc metallopeptidase STE24) is a nuclear membrane zinc metalloprotease. It possesses seven transmembrane α-helical barrel shaped structure with a large, water-filled, intramembrane chamber across periphery. It also has a zinc metalloprotease domain with the catalytic site facing into the intramembrane chamber. It endoproteolytically cleaves prelamin A, a component of the nuclear lamina intermediate filaments. The absence of this cleaving activity leads to the prelamin A accumulation in cells that further causes laminopathies with premature aging disease progeria and metabolic disorders. Mutation in ZMPSTE24 also causes a rare autosomal recessive progeroid syndrome, mandibuloacral dysplasia (MAD).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70780

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jemima Barrowman et al.
Biological chemistry, 390(8), 761-773 (2009-05-21)
ZMPSTE24 is an integral membrane zinc metalloprotease originally discovered in yeast as an enzyme (called Ste24p) required for maturation of the mating pheromone a-factor. Surprisingly, ZMPSTE24 has recently emerged as a key protease involved in human progeroid disorders. ZMPSTE24 has
Y Miyoshi et al.
Clinical genetics, 73(6), 535-544 (2008-04-26)
Mandibuloacral dysplasia (MAD) is a rare autosomal recessive progeroid syndrome, characterized by mandibular hypoplasia, acroosteolysis affecting distal phalanges and clavicles, delayed closure of the cranial sutures, atrophic skin, and lipodystrophy. Recently, mutations in lamin A/C (LMNA) and zinc metalloprotease (ZMPSTE24)
Andrew Quigley et al.
Science (New York, N.Y.), 339(6127), 1604-1607 (2013-03-30)
Mutations in the nuclear membrane zinc metalloprotease ZMPSTE24 lead to diseases of lamin processing (laminopathies), such as the premature aging disease progeria and metabolic disorders. ZMPSTE24 processes prelamin A, a component of the nuclear lamina intermediate filaments, by cleaving it

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service