Skip to Content
Merck
All Photos(4)

Key Documents

HPA006429

Sigma-Aldrich

Anti-TAF7 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-TAF(II)55 antibody produced in rabbit, Anti-TAFII-55 antibody produced in rabbit, Anti-TAFII55 antibody produced in rabbit, Anti-Transcription initiation factor TFIID 55 kDa subunit antibody produced in rabbit, Anti-Transcription initiation factor TFIID subunit 7 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Check Cart for Availability


Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$1,040.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

ISSPGMSGHRQGHDSLEHDELREIFNDLSSSSEDEDETQHQDEEDINIIDTEEDLERQLQDKLNESDEQHQENEGTNQLVMGIQKQIDNMKGKLQETQDRAKRQEDLIMKVENLALKN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TAF7(6879)

General description

TAF7 (TATA-box binding protein associated factor 7) gene is localized to human chromosome 5q31. It is a part of the multi-subunit transcription factor TFIID. It was initially recognized as TBP-associated factor (TAFII) of TFIID, which is specific for RNA polymerase II.

Immunogen

Transcription initiation factor TFIID subunit 7 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

TAF7 (TATA-box binding protein associated factor 7) acts as a transcriptional repressor, where it binds to TAF1, and prevents its acetyl transferase activity. This activity is necessary for the basal transcription of MHC (major histocompatibility class) I. It acts as a regulator of check-point and prevents premature initiation of transcription until the preinitiation complex (PIC) is assembled completely. It interacts with multiple transcription factors such as, Sp1 (specificity protein), YY1 (Yin Yang 1), USF (upstream transcription factor 1), CTF (C-terminal fragment), adenovirus E1A, and HIV-1 (human immunodeficiency virus) Tat (trans-activator of transcription). It is also involved in the activation of thyroid hormone and vitamin D3 receptor in a ligand-independent fashion. It might also be involved in the processing of the 3′ end of mRNA.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70992

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Zhou et al.
The Journal of biological chemistry, 276(27), 25503-25511 (2001-05-08)
Human TAF(II)55 (hTAF(II)55) is a component of the multisubunit general transcription factor TFIID and has been shown to mediate the functions of many transcriptional activators via direct protein-protein interactions. To uncover the regulatory properties of the general transcription machinery, we
Anthony R Dallosso et al.
PLoS genetics, 5(11), e1000745-e1000745 (2009-12-04)
Wilms' tumour (WT) is a pediatric tumor of the kidney that arises via failure of the fetal developmental program. The absence of identifiable mutations in the majority of WTs suggests the frequent involvement of epigenetic aberrations in WT. We therefore
Anne Gegonne et al.
Proceedings of the National Academy of Sciences of the United States of America, 103(3), 602-607 (2006-01-13)
Transcription consists of a series of highly regulated steps: assembly of a preinitiation complex (PIC) at the promoter nucleated by TFIID, followed by initiation, elongation, and termination. The present study has focused on the role of the TFIID component, TAF7

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service