Skip to Content
Merck
All Photos(3)

Key Documents

HPA006034

Sigma-Aldrich

Anti-PPP2R5E antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-PP2A, B subunit, B′ ε-isoform antibody produced in rabbit, Anti-PP2A, B subunit, B56 ε-isoform antibody produced in rabbit, Anti-PP2A, B subunit, PR61 ε-isoform antibody produced in rabbit, Anti-PP2A, B subunit, R5 ε-isoform antibody produced in rabbit, Anti-Serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit ε-isoform antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Check Cart for Availability


Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$1,040.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

MSSAPTTPPSVDKVDGFSRKSVRKARQKRSQSSSQFRSQGKPIELTPLPLLKDVPSSEQPELFLKKLQQCCVIFDFMDTLSDLKMKEYKRSTLNELVDYITISRGCLTEQTYPEVVRMVSCNIFRTLPPSDSNEFDPEEDEPTLEASWPH

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PPP2R5E(5529)

Immunogen

Serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit ε-isoform recombinant protein epitope signature tag (PrEST)

Application

Anti-PPP2R5E antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit epsilon isoform is an enzyme encoded by the PPP2R5E gene in humans. This gene belongs to the phosphatase 2A regulatory subunit B family. It acts as a tumor suppressor in human cancer. Down-regulation of this gene by miR-23a, induces cell growth inhibition and apoptosis in gastric cancer cells. Variants in this gene can influence human cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70418

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lukasz F Grochola et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 15(19), 6301-6308 (2009-09-24)
A regulated p53-dependent stress response is crucial in suppressing tumor formation and mediating the response to commonly used cancer therapeutics. However, little is known about the human, inherited genetics of this important signaling pathway. Studies of human genetic variants in
Xue Liu et al.
FEBS letters, 588(17), 3160-3169 (2014-07-06)
PPP2R5E belongs to the phosphatase 2A regulatory subunit B family and acts as a tumor suppressor in human cancer. However, the role of PPP2R5E in the tumorigenesis of gastric cancer is unclear. Here, we declare that PPP2R5E is downregulated by
Prajakta Varadkar et al.
Cell cycle (Georgetown, Tex.), 16(12), 1210-1219 (2017-06-01)
The Spindle Assembly Checkpoint (SAC) is part of a complex feedback system designed to ensure that cells do not proceed through mitosis unless all chromosomal kinetochores have attached to spindle microtubules. The formation of the kinetochore complex and the implementation
Sean T DeGrande et al.
The Journal of biological chemistry, 288(2), 1032-1046 (2012-12-04)
Kinase/phosphatase balance governs cardiac excitability in health and disease. Although detailed mechanisms for cardiac kinase regulation are established, far less is known regarding cardiac protein phosphatase 2A (PP2A) regulation. This is largely due to the complexity of the PP2A holoenzyme

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service