Skip to Content
Merck
All Photos(8)

Key Documents

HPA005729

Sigma-Aldrich

Anti-GALT antibody produced in rabbit

enhanced validation

Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Gal-1-P uridylyltransferase, Anti-UDP-glucose-hexose-1-phosphate uridylyltransferase

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$952.00

$952.00


Check Cart for Availability


Select a Size

Change View
100 μL
$952.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

$952.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

SEHWLVLVPFWATWPYQTLLLPRRHVRRLPELTPAERDDLASIMKKLLTKYDNLFETSFPYSMGWHGAPTGSEAGANWNHWQLHAHYYPPLLRSATVRKFMVGYEMLAQAQRDLTPEQAAERLRALPEVHYHLGQKDRETA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GALT(2592)

Looking for similar products? Visit Product Comparison Guide

Specificity

Note: The Ensemble Gene ID has changed from ENSG00000137070 in the 46:36 release of the database to ENSG00000213930 in the 48:36 release of the database.

Immunogen

Galactose-1-phosphate uridylyltransferase recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Galactose-1-phosphate uridylyltransferase (also known as GALT) is an enzyme essential for converting ingested galactose to glucose. Deficiency of this gene causes classic galactosemia, an autosomal recessive metabolic disorder. It may lead to toxic accumulation of galactose and derived metabolites. Mutation in this gene is ethnic-specific and galactosemia is a heterogeneous disorder at the molecular level.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86838

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rihwa Choi et al.
BMC medical genetics, 15, 94-94 (2014-08-16)
Classic galactosemia (OMIM #230400) is an autosomal recessive metabolic disorder caused by a deficiency of the galactose-1-phosphate uridyltransferase (GALT, EC2.7.7.12) protein due to mutations in the GALT gene. The aim of this study was to provide a comprehensive and updated
E Viggiano et al.
Gene, 559(2), 112-118 (2015-01-17)
Classical galactosemia is an autosomal recessive inborn error of metabolism due to mutations of the GALT gene leading to toxic accumulation of galactose and derived metabolites. With the benefit of early diagnosis by neonatal screening and early therapy, the acute
Sylvia C Estrada et al.
The Kobe journal of medical sciences, 59(3), E106-E111 (2013-09-21)
Classic galactosemia is an inherited metabolic disorder due to mutations in the galactose-1-phosphate uridyltransferase (GALT) gene. This study describes the results of the GALT gene analysis of four unrelated Filipino patients with Classic Galactosemia. DNA extracted from dried blood spots
Rıza Köksal Özgül et al.
Journal of human genetics, 58(10), 675-678 (2013-08-09)
Classical galactosemia is an inherited recessive disorder of galactose metabolism caused by deficiency of the enzyme galactose-1-phosphate uridyl transferase (GALT), which is caused by mutations in the GALT gene. In this study, 56 Turkish patients diagnosed with galactosemia were screened
Maria Luisa Barretta et al.
Nature communications, 7, 11727-11727 (2016-06-01)
The Golgi apparatus is composed of stacks of cisternae laterally connected by tubules to form a ribbon-like structure. At the onset of mitosis, the Golgi ribbon is broken down into discrete stacks, which then undergo further fragmentation. This ribbon cleavage

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service