Skip to Content
Merck
All Photos(3)

Key Documents

HPA005689

Sigma-Aldrich

Anti-BLM antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Bloom syndrome protein, Anti-DNA helicase, RecQ-like type 2, Anti-RecQ protein-like 3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$952.00

$952.00


Check Cart for Availability


Select a Size

Change View
100 μL
$952.00

About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$952.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

STLKDLDTSDRKEDVLSTSKDLLSKPEKMSMQELNPETSTDCDARQISLQQQLIHVMEHICKLIDTIPDDKLKLLDCGNELLQQRNIRRKLLTEVDFNKSDASLLGSLWRYRPDSLDGPMEGDSCPTGNSMKELNFSHLPSNSV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BLM(641)

General description

Bloom syndrome protein (BLM) is a DNA helicase belonging to the RecQ family, having 1417 amino acids. The gene encoding it is present on chromosome 15. BLM contains an amino-terminal and a carboxy-terminal region which is composed predominantly of charged polar amino acid residues.

Immunogen

Bloom syndrome protein recombinant protein epitope signature tag (PrEST)

Application

Anti-BLM antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige. Anti-BLM antibody produced in rabbit is also suitable for immunoprecipitation.[1]
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)

Biochem/physiol Actions

Bloom syndrome protein (BLM) plays an important role in the maintenance of genomic stability in somatic cells. It functions in the complex processes of DNA replication where it interacts with topoisomerase II. BLM is also a part of a DNA surveillance mechanism (BRCA1-associated) operating during S phase and is found in the nucleolus of S Phase cells, interacting with a protein WRN. It binds to p53, topoisomerase IIIa, RAD5 (a DNA repair protein) and is thus associated with several aspects of DNA metabolism like DNA recombination, replication, and repair.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST78241

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cayetano von Kobbe et al.
The Journal of biological chemistry, 277(24), 22035-22044 (2002-03-29)
The RecQ helicase family comprises a conserved group of proteins implicated in several aspects of DNA metabolism. Three of the family members are defective in heritable diseases characterized by abnormal growth, premature aging, and predisposition to malignancies. These include the
Victoria Valinluck Lao et al.
Translational oncology, 6(4), 458-469 (2013-08-03)
Deregulation of DNA repair enzymes occurs in cancers and may create a susceptibility to chemotherapy. Expression levels of DNA repair enzymes have been shown to predict the responsiveness of cancers to certain chemotherapeutic agents. The RECQ helicases repair damaged DNA
N A Ellis et al.
Cell, 83(4), 655-666 (1995-11-17)
The Bloom's syndrome (BS) gene, BLM, plays an important role in the maintenance of genomic stability in somatic cells. A candidate for BLM was identified by direct selection of a cDNA derived from a 250 kb segment of the genome
V Yankiwski et al.
Proceedings of the National Academy of Sciences of the United States of America, 97(10), 5214-5219 (2000-04-26)
Bloom syndrome (BS) is a rare cancer-predisposing disorder in which the cells of affected persons have a high frequency of somatic mutation and genomic instability. BLM, the protein altered in BS, is a RecQ DNA helicase. This report shows that
Suruchika Chandra et al.
Journal of cell science, 126(Pt 16), 3782-3795 (2013-06-12)
The spectrum of tumors that arise owing to the overexpression of c-Myc and loss of BLM is very similar. Hence, it was hypothesized that the presence of BLM negatively regulates c-Myc functions. By using multiple isogenic cell lines, we observed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service