Skip to Content
Merck
All Photos(7)

Key Documents

HPA005554

Sigma-Aldrich

Anti-ARSA antibody produced in rabbit

enhanced validation

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ASA antibody produced in rabbit, Anti-Arylsulfatase A precursor antibody produced in rabbit, Anti-Cerebroside-sulfatase antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$952.00

$952.00


Check Cart for Availability


Select a Size

Change View
100 μL
$952.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$952.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:500-1:1000

immunogen sequence

LLLGTGKSPRQSLFFYPSYPDEVRGVFAVRTGKYKAHFFTQGSAHSDTTADPACHASSSLTAHEPPLLYDLSKDPGENYNLLGGVAGATPEVLQALKQLQLLKAQLDAAVTFGPSQVARGEDPALQICCHPGCTPRPACCHCP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ARSA(410)

General description

ARSA (arylsulfatase A) is a lysosomal sulfatase. This gene maps to human chromosome 22q13. This protein exists as a dimer and has a molecular weight of 107kDa. It is an acidic glycoprotein. It consists of 507 amino acid residues. Placental arylsulfatase A contains a putative 18 amino acid long signal peptide, preceding its N- terminal. ARSA is predicted to have three N-glycosylation sites.

Immunogen

Arylsulfatase A precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-ARSA antibody is suitable for co-immunoprecipitation, co-localization and flow cytometry.
Anti-ARSA antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

ARSA (arylsulfatase A) requires post-translational modification to become catalytically active. This involves the oxidation of the -CH2SH group of a cysteine residue to aldehyde. Lack of this modification results in inactive enzyme, which causes the lysosomal storage disorder called multiple sulfatase deficiency. Complete or partial inactivation of this gene leads to Metachromatic leukodystrophy (MLD), which is a lysosomal storage disease. It is characterized by the accumulation of cerebroside sulfate in lysosomes. ARSA pseudo-deficiency is caused in individuals who are homozygous for the allele, which results in significant loss of ARSA activity. However, the activity is sufficient for normal cerebroside catabolism and results in clinically healthy phenotype.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73374

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C DeLuca et al.
Proceedings of the National Academy of Sciences of the United States of America, 76(4), 1957-1961 (1979-04-01)
Genetics of human lysosomal arylsulfatases A and B (aryl-sulfate sulfohydrolase, EC 3.1.6.1), associated with childhood disease, has been studied with human-rodent somatic cell hybrids. Deficiency of arylsulfatase A (ARS(A)) in humans results in a progressive neurodegenerative disease, metachromatic leukodystrophy. Deficiency
V Gieselmann et al.
American journal of human genetics, 49(2), 407-413 (1991-08-01)
We identified a patient suffering from late infantile metachromatic leukodystrophy who genetically seemed to be homozygous for the mutations signifying the arylsulfatase A pseudodeficiency allele. Homozygosity for the pseudodeficiency allele is associated with low arylsulfatase A activity but does not
G Lukatela et al.
Biochemistry, 37(11), 3654-3664 (1998-04-02)
Human lysosomal arylsulfatase A (ASA) is a prototype member of the sulfatase family. These enzymes require the posttranslational oxidation of the -CH2SH group of a conserved cysteine to an aldehyde, yielding a formylglycine. Without this modification sulfatases are catalytically inactive
Ruben J Boado et al.
Biotechnology and bioengineering, 110(5), 1456-1465 (2012-11-30)
Metachromatic leukodystrophy (MLD) is a lysosomal storage disorder of the brain caused by mutations in the gene encoding the lysosomal sulfatase, arylsulfatase A (ASA). It is not possible to treat the brain in MLD with recombinant ASA, because the enzyme
Agnieszka Ługowska et al.
PloS one, 6(6), e20218-e20218 (2011-06-23)
Metachromatic leukodystrophy (MLD) is a severe neurometabolic disease caused mainly by deficiency of arylsulfatase A encoded by the ARSA gene. Based on epidemiological surveys the incidence of MLD per 100,000 live births varied from 0.6 to 2.5. Our purpose was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service