Skip to Content
Merck
All Photos(6)

Key Documents

HPA004827

Sigma-Aldrich

Anti-USP13 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Deubiquitinating enzyme 13 antibody produced in rabbit, Anti-ISOT-3 antibody produced in rabbit, Anti-Isopeptidase T-3 antibody produced in rabbit, Anti-Ubiquitin carboxyl-terminal hydrolase 13 antibody produced in rabbit, Anti-Ubiquitin thioesterase 13 antibody produced in rabbit, Anti-Ubiquitin-specific-processing protease 13 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

RKAVYFTGNMGAEVAFNWIIVHMEEPDFAEPLTMPGYGGAASAGASVFGASGLDNQPPEEIVAIITSMGFQRNQAIQALRATNNNLERALDWIFSHPEFEEDSDFVIEMENNANANIISEAKPEGPRVKDGSGTYELFAFISH

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... USP13(8975)

Immunogen

Ubiquitin carboxyl-terminal hydrolase 13 recombinant protein epitope signature tag (PrEST)

Application

Anti-USP13 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

USP13 (Ubiquitin specific peptidase 13) is a deubiquitinating enzyme involved in the deubiquitination of microphthalmia-associated transcription factor (MITF). It is highly expressed in ovary, testes and lesser extent in other tissues. MITF is a key component of melanocyte development. USP13 generates stable and upregulated MITF protein through the deubiquitination pathway. Therefore, suppression of USP13 can cause dramatic loss of MITF protein. Studies have suggested that USP13 may have therapeutic aspects in melanoma cells. It has been reported that USP13 may play an important role in the JAK-STAT signaling pathway. It regulates type I and type II interferon signaling by deubiquitinating and stabilizing STAT1.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86788

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiansi Zhao et al.
Nature communications, 2, 414-414 (2011-08-04)
The microphthalmia-associated transcription factor (MITF) is essential for melanocyte development. Mutation-induced MAPK pathway activation is common in melanoma and induces MITF phosphorylation, ubiquitination, and proteolysis. Little is known about the enzymes involved in MITF ubiquitination/deubiquitination. Here we report the identification
Hoi-Hung Cheung et al.
British journal of cancer, 114(2), 230-236 (2015-12-02)
Testicular embryonal carcinoma (EC) is a major subtype of non-seminomatous germ cell tumours in males. Embryonal carcinomas are pluripotent, undifferentiated germ cell tumours believed to originate from primordial germ cells. Epigenetic changes during testicular EC tumorigenesis require better elucidation. To
Qiao Cheng et al.
European journal of immunology, 51(2), 296-310 (2020-07-17)
Viral infection is a serious threat to both normal population and clinical patients. STAT1 plays central roles in host defense against viral infection. How STAT1 protein maintains stable in different conditions remains largely unknown. Here, we identified BRCC36 as a
K M Timms et al.
Gene, 217(1-2), 101-106 (1998-12-05)
A novel Isopeptidase T gene (ISOT-3) has been identified on human mosome 3q26.2--q26.3. gene shows 67.3% nucleotide identity and 54.8% amino acid identity to n Isopeptidase (ISOT-1). Northern blot analysis has shown that ISOT-3 is highly essed in ovary and
Hom-Ming Yeh et al.
Journal of immunology (Baltimore, Md. : 1950), 191(6), 3328-3336 (2013-08-14)
The IFN immune system comprises type I, II, and III IFNs, signals through the JAK-STAT pathway, and plays central roles in host defense against viral infection. Posttranslational modifications such as ubiquitination regulate diverse molecules in the IFN pathway. To search

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service