Skip to Content
Merck
All Photos(2)

Key Documents

HPA004184

Sigma-Aldrich

Anti-LIMCH1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-NP_055803.1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Check Cart for Availability


Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$1,040.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

HTEEVKLIVTCNMRAQESEPVEGGLRKVPDLHKDDLAQQRIQGSLAPHREPPSFITLSNITEADLETWERLKVSEKARDGDVQHICASEPSPEIKAETAIRDDFANRKARASKKASSPRQKFVHFGPVTELDQQKW

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LIMCH1(22998)

General description

LIMCH1 belongs to the human mosaic domain superfamily. It is multidomain molecule composed of calponin homology (CH) domain that acts as actin binding domain and the conserved cysteine rich double Zn finger possessing Lim domain. Both the domain exists as a singlet. It has reported that these proteins may exist in several duplex and quadruplet CH containing forms. It may involve in the cytoskeletal and signaling pathways.

Immunogen

LIM and calponin homology domains-containing protein 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86758

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Felix Friedberg
Molecular biology reports, 37(4), 1707-1716 (2009-07-01)
In this paper, the inventory presented for singlet CH (calponin homology/actin binding) domain containing human multidomain proteins is extended to several duplex and one quadruplet CH containing forms. Invariably, the duplexes are located at the begin of the molecules. The
T Stradal et al.
FEBS letters, 431(2), 134-137 (1998-08-26)
A sequence motif of about 100 amino acids, termed the 'calponin homology domain' has been suggested to confer actin binding to a variety of cytoskeletal and signalling molecules. Here we analyse and compare the sequences of all calponin homology domain-containing
Felix Friedberg
Molecular biology reports, 36(5), 1059-1081 (2008-06-17)
In this paper we examine (restricted to homo sapiens) the products resulting from gene duplication and the subsequent alternative splicing for the members of a multidomain group of proteins which possess the evolutionary conserved calponin homology CH domain, i.e. an

Questions

  1. We bought this LIMCH1 Ab. Can this LIMCH1 Ab used for the immunoprecipitation using human trabecular meshwork cells?

    1 answer
    1. This product is the Prestige antibody from Atlas Antibodies, and all quality testing is performed by Atlas Antibodies. The antibody has been validated through immunohistochemistry tissue arrays of 44 normal human tissues and 20 common cancer type tissues, as well as a protein array of 364 human recombinant protein fragments. It is recommended for immunohistochemistry applications only. The performance of this antibody in immunoprecipitation assays is unknown, and it is not recommended for such use. If use in immunoprecipitation is desired, empirical determination of its suitability is necessary.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service