Skip to Content
Merck
All Photos(6)

Key Documents

HPA003948

Sigma-Aldrich

Anti-ITIH4 antibody produced in rabbit

enhanced validation

Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-GP120, Anti-IHRP, Anti-ITI heavy chain H4, Anti-Inter-alpha-inhibitor heavy chain 4, Anti-Inter-alpha-trypsin inhibitor family heavy chain-related protein, Anti-MUSTN1 antibody produced in rabbit, Anti-PK-120, Anti-Plasma kallikrein sensitive glycoprotein 120, Anti-Transmembrane protein 110 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Check Cart for Availability


Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$1,040.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

LAYSFVTPLTSMVVTKPDDQEQSQVAEKPMEGESRNRNVHSAGAAGSRMNFRPGVLSSRQLGLPGPPDVPDHAAYHPFRRLAILPASAPPATSNPDPAVSRVMNMKIEETTMTTQTPA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ITIH4(3700)

Looking for similar products? Visit Product Comparison Guide

Immunogen

Transmembrane protein 110 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Inter-α-trypsin inhibitor heavy chain H4 is a protein encoded by the ITIH4 gene in humans. The protein is a 120kDa acute-phase glycoprotein produced primarily in the liver, secreted into the blood, and identified in serum. It is involved in liver development and stabilization of the extracellular matrix (ECM) and its expression is found to be altered in liver disease.[1] This gene is an anti-inflammatory protein and is a substrate for plasma kallikrein (PK). It shows significant sequence identities to heavy chains (HCs) of the inter-α-trypsin inhibitor (ITI) superfamily. Its expression may be used as a biomarker that may facilitate the development of novel diagnostic and therapeutic tools.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST84483

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kevin Brown Chandler et al.
Journal of proteome research, 13(7), 3314-3329 (2014-06-03)
Inter-alpha-trypsin inhibitor heavy chain H4 (ITIH4) is a 120 kDa acute-phase glycoprotein produced primarily in the liver, secreted into the blood, and identified in serum. ITIH4 is involved in liver development and stabilization of the extracellular matrix (ECM), and its
Myung-Sun Kim et al.
Molecular bioSystems, 7(5), 1430-1440 (2011-02-19)
Recurrent pregnancy loss (RPL) is defined as at least three pregnancy losses in series prior to the 20-28 weeks of pregnancy. There are several etiological factors associated with immunology, anatomy, endocrinology, genetic, infection, chromosomal abnormalities, and environmental factors contributing to
H Nishimura et al.
FEBS letters, 357(2), 207-211 (1995-01-03)
PK-120 is a substrate for plasma kallikrein (PK), recently purified from human plasma. Here we have established the cDNA sequence for human PK-120 mRNA. The deduced amino sequence of PK-120 revealed that it consists of 902 amino acid residues with
Rajpal S Kashyap et al.
Clinica chimica acta; international journal of clinical chemistry, 402(1-2), 160-163 (2009-03-06)
Inflammation and inflammatory markers play an important role in acute ischemic stroke (AIS). In an earlier study, we discovered a 120-kDa protein that was highly expressed in healthy participants, but either was not expressed or was barely expressed in AIS

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service