Skip to Content
Merck
All Photos(7)

Key Documents

HPA003936

Sigma-Aldrich

Anti-CCDC134 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Coiled-coil domain-containing protein 134 precursor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Check Cart for Availability


Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$1,040.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

LLALKNLAQLNDIHQQYKILDVMLKGLFKVLEDSRTVLTAADVLPDGPFPQDEKLKDAFSHVVENTAFFGDVVLRFPRIVHYYFDHNSNWNLLIRWGISFCNQTGVFNQGPHSPILSLMAQEL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CCDC134(79879)

Immunogen

Coiled-coil domain-containing protein 134 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CCDC134 (coiled coil domain containing 134) is a novel classical secretory protein and is a member of the secretory family. It acts as an inhibitor of Raf-1/MEK/ERK and JNK/ SAPK pathways. The gene encodes a secretory protein that can inhibit the MAPK pathway as a novel human MAPK-regulating protein. It is widely expressed in normal adult tissues, tumor tissues and cell lines. It may be a candidate biomarker for malignant transformation. It plays a role in regulation of cell migration and invasion and may act as a therapeutic target for gastric cancer. It may act as an immune cytokine and may serve as a member of the γ(c) cytokine family.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73553

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Huang et al.
Cellular and molecular life sciences : CMLS, 65(2), 338-349 (2007-12-19)
In this study, we report a novel gene, CCDC134 (coiled-coil domain containing 134), that encodes a secretory protein that can inhibit the MAPK pathway as a novel human MAPK-regulating protein. The CCDC134 mRNA contains 1280 nucleotides, encoding a protein of
Jialing Zhong et al.
Molecular and cellular biochemistry, 372(1-2), 1-8 (2012-10-17)
CCDC134 (coiled coil domain containing 134), a novel secretory protein, acts as an inhibitor of Erk1/2 and JNK/SAPK pathways. However, the role of CCDC134 in cancer development is still lacking. In this study, we found that CCDC134 expression significantly reduced
Jing Huang et al.
Cancer research, 74(20), 5734-5745 (2014-08-16)
CCDC134 is a poorly characterized secreted protein that may act as an immune cytokine. Here, we show that CCDC134 is differentially expressed on resting and activated immune cells and that it promotes CD8(+) T-cell activation, proliferation, and cytotoxicity by augmenting

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service