Skip to Content
Merck
All Photos(3)

Key Documents

HPA003897

Sigma-Aldrich

Anti-EPS8 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

EPS8 Antibody - Anti-EPS8 antibody produced in rabbit, Eps8 Antibody, Anti-Epidermal growth factor receptor kinase substrate 8

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$952.00

$952.00


Check Cart for Availability


Select a Size

Change View
100 μL
$952.00

About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

$952.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

PKEQFIPPYVPRFRNGWEPPMLNFMGATMEQDLYQLAESVANVAEHQRKQEIKRLSTEHSSVSEYHPADGYAFSSNIYTRGSHLDQGEAAVAFKPTSNRHIDRNYEPLKTQPKKYAKSKYDFVARNNSELSVLKDDILEILDD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EPS8(2059)

Immunogen

Epidermal growth factor receptor kinase substrate 8 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Epidermal growth factor receptor kinase substrate 8 is an enzyme encoded by the EPS8 gene in humans and is mapped to chromosome 12q23-q24. It is a novel proto-oncogene related to the tumorigenesis, proliferation, metastasis, chemo-resistance and prognosis of various human solid cancers. The gene is a key regulator in cell signaling, fibroblast growth factor receptor (FGFR) signaling and receptor trafficking. It is a substrate of the epidermal growth factor receptor and its overexpression leads to increased mitogenic signaling and malignant transformation. It is overexpressed in human gliomas, and affects glioma cell growth possibly by regulating extracellular signal-regulated protein kinase (ERK) and Akt/β-catenin signaling.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86670

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Wang et al.
Clinical laboratory, 59(11-12), 1261-1269 (2014-01-15)
Eps8 is a novel proto-oncogene related to the tumorigenesis, proliferation, metastasis, chemo-resistance, and prognosis of many human solid cancers. However, the function of Eps8 in acute myeloid leukemia (AML) is still unclear. Thus, this study aims to develop a real-time
Giulio Auciello et al.
Journal of cell science, 126(Pt 2), 613-624 (2012-12-04)
Fibroblast growth factor receptors (FGFRs) mediate a wide spectrum of cellular responses that are crucial for development and wound healing. However, aberrant FGFR activity leads to cancer. Activated growth factor receptors undergo stimulated endocytosis, but can continue to signal along
Haruhi Fukuhisa et al.
Journal of human genetics, 64(6), 521-534 (2019-03-13)
Our ongoing analyses identifying dysregulated microRNAs (miRNAs) and their controlled target RNAs have shed light on novel oncogenic pathways in pancreatic ductal adenocarcinoma (PDAC). The PDAC miRNA signature obtained by RNA sequencing showed that both strands of pre-miR-130b (miR-130b-5p, the
W T Wong et al.
Oncogene, 9(10), 3057-3061 (1994-10-01)
We have previously isolated the coding sequence for a novel substrate for tyrosine kinases, eps8, from NIH3T3 fibroblasts. Eps8 was phosphorylated in vivo by several receptor tyrosine kinases (RTKs) and, upon overexpression, was able to enhance EGFR-mediated mitogenic signaling in
Debbie L Cunningham et al.
PloS one, 8(4), e61513-e61513 (2013-04-30)
Eps8 is involved in both cell signalling and receptor trafficking. It is a known phosphorylation substrate for two proteins involved in the fibroblast growth factor receptor (FGFR) signalling pathway: the receptor itself and Src. Here we report a differential proteomic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service