Skip to Content
Merck
All Photos(2)

Documents

HPA003656

Sigma-Aldrich

Anti-PNPLA4 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-DXS1283E, Anti-GS2, Anti-iPLA2eta

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

QFTYKFAEEIRRQSFGAVTPGYDFMARLRSGMESILPPSAHELAQNRLHVSITNAKTRENHLVSTFSSREDLIKVLLASSFVPIYAGLKLVEYKGQKWVDGGLTNALPILPVGRTVTISPFSGRLDISPQDKGQLD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PNPLA4(8228)

Immunogen

Patatin-like phospholipase domain-containing protein 4 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PNPLA4 belongs to patatin-like family of phospholipases (PNPLA) and is the smallest member. The gene is expressed in all human tissues such as heart, brain, placenta, lung, liver, muscle, kidney, pancreas and spleen. It hydrolyzes retinylesters (RE) in cell homogenates, promotes RE accumulation. It may act as a catalyst or as a regulatory protein that enhances RE formation catalyzed by other acyl transferases. In humans, it also catalyzes keratinocyte retinylester and triglyceride hydrolysis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74329

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jay G Gao et al.
Biochemical and biophysical research communications, 360(2), 501-506 (2007-07-03)
We have previously shown that human GS2 (hGS2) catalyzes keratinocyte retinylester and triglyceride hydrolysis. hGS2 and its rat orthologue, rGS2, are 80% homologous and share a proline insertion at residue 56 and a C-terminal truncation compared to the hGS2 paralogues.
Jay G Gao et al.
Molecular genetics and metabolism, 96(4), 253-260 (2009-02-03)
GS2 (PNPLA4; iPLAeta) is the smallest member of the patatin-like family of phospholipases (PNPLA). It was initially identified by its ability to hydrolyze retinylesters (RE) in cell homogenates, and was later found to esterify retinol using a variety of acyl
W C Lee et al.
Genomics, 22(2), 372-376 (1994-07-15)
A gene, GS2 (DXS1283E), was isolated from a CpG island located approximately midway between the steroid sulfatase (STS) and the Kallmann syndrome (KAL1) loci on the distal short arm of the human X chromosome. DNA sequencing of a GS2 cDNA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service