Skip to Content
Merck
All Photos(3)

Key Documents

HPA003635

Sigma-Aldrich

Anti-MAD1L1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-HsMAD1, Anti-MAD1, Anti-MAD1-like 1, Anti-Mitotic arrest deficient-like protein 1, Anti-Mitotic checkpoint MAD1 protein-homolog, Anti-Mitotic spindle assembly checkpoint protein, Anti-Tax-binding protein 181, Anti-hMAD1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$952.00

$952.00


Estimated to ship on26 May 2025



Select a Size

Change View
100 μL
$952.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$952.00


Estimated to ship on26 May 2025


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200-1:500
western blot: 0.04-0.4 μg/mL

immunogen sequence

TKVLHMSLNPTSVARQRLREDHSQLQAECERLRGLLRAMERGGTVPADLEAAAASLPSSKEVAELKKQVESAELKNQRLKEVFQTKIQEFRKACYTLTGYQIDITTENQYRLTSLYAEHPGDCLIFKATSPSGSKMQLLEIRRAPPLSNN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAD1L1(8379)

Immunogen

Mitotic spindle assembly checkpoint protein recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Mitotic spindle assembly checkpoint protein MAD1 is a protein encoded by the MAD1L1 gene in humans and is mapped to human chromosome 7p22. It is essential for mitotic arrest and has an additional function in promoting the checkpoint. Alteration in its expression is associated with chromosomal instability and might be used as a prognostic biomarker for breast cancer. Its expression in nuclei is a predictive biomarker of contraindication to pacilitaxel treatment in breast cancer. The protein may act as a potential target for sensitizing cancer cells to redox-cycling drugs.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86432

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qian Sun et al.
Breast cancer research and treatment, 140(2), 323-330 (2013-07-19)
Aneuploidy is a characteristic of human cancers, and recent studies have suggested that defects of mitotic checkpoints play a role in carcinogenesis. Mitotic Arrest Deficient-Like 1 (MAD1L1), whose altered expression is associated with chromosomal instability, is a checkpoint gene. We
Kumsun Cho et al.
Free radical biology & medicine, 60, 201-210 (2013-03-06)
Cancer cells acquire resistance to chemotherapy under hypoxia, which is mainly driven by the transcription factor HIF (hypoxia-inducible factor). Yet, it is uncertain which molecules mediate such resistance. While profiling gene expression in colon cancer cells, we found that Mad1
Thomas Kruse et al.
EMBO reports, 15(3), 282-290 (2014-01-31)
The spindle assembly checkpoint (SAC) ensures accurate chromosome segregation by delaying entry into anaphase until all sister chromatids have become bi-oriented. A key component of the SAC is the Mad2 protein, which can adopt either an inactive open (O-Mad2) or
Mitotic checkpoint locus MAD1L1 maps to human chromosome 7p22 and mouse chromosome 5.
D Y Jin et al.
Genomics, 55(3), 363-364 (1999-03-02)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service