Skip to Content
Merck
All Photos(5)

Key Documents

HPA003405

Sigma-Aldrich

Anti-IVNS1ABP antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-influenza virus NS1A binding protein antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Estimated to ship on27 May 2025



Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$1,040.00


Estimated to ship on27 May 2025


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

QDELIEKPMSPMQYARSGLGTAEMNGKLIAAGGYNREECLRTVECYNPHTDHWSFLAPMRTPRARFQMAVLMGQLYVVGGSNGHSDDLSCGEMYDSNIDDWIPVPELRTNRCNAGVCALNGKLYIVGGSD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Influenza virus NS1A-binding protein is a protein encoded by the IVNS1ABP gene in humans. It is a 70kDa protein that interacts with the nonstructural NS1 protein of the influenza A virus. It consists an N-terminal BTB/POZ domain and five kelch-like tandem repeat elements of approximately 50 amino acids. It is found in the nucleus (nucleoplasm). The gene IVNS1ABP is mapped to human chromosome 1q25.1-q31.1.

Immunogen

influenza virus NS1A binding protein recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Influenza virus NS1A-binding protein is found to interact with actin in human cells and plays an important function in controlling cell proliferation. The gene also helps in regulating the concentration of functional aryl hydrocarbon receptor (AHR; it is a ligand-activated transcription factor with important roles in metabolic adaptation, dioxin toxicology and vascular development) in mammalian cells.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86649

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barbara Bennani-Baiti et al.
Gene, 491(2), 103-109 (2011-10-25)
Several gene databases, including heavily used ones such as the National Center for Biotechnology Information (NCBI) database, erroneously assign, on occasion, literature references to genes or proteins. These mistakes are mostly due to an overlap in gene aliases, whereby two
Elizabeth E Dunham et al.
Molecular pharmacology, 70(1), 8-15 (2006-04-04)
The aryl hydrocarbon receptor (AHR) is a ligand-activated transcription factor with important roles in metabolic adaptation, dioxin toxicology, and vascular development. To understand the details of this signal transduction pathway, we have used the yeast two-hybrid system to identify proteins
Giovanni Perconti et al.
Biochimica et biophysica acta, 1773(12), 1774-1785 (2007-11-13)
Alpha-enolase is a key glycolytic enzyme that plays a functional role in several physiological processes depending on the cellular localization. The enzyme is mainly localized in the cytoplasm whereas an alternative translated form, named MBP-1, is predominantly nuclear. The MBP-1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service