Skip to Content
Merck
All Photos(6)

Key Documents

HPA003218

Sigma-Aldrich

Anti-DOCK8 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Dedicator of cytokinesis protein 8 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.4 μg/mL
immunohistochemistry: 1:500- 1:1000

immunogen sequence

AIANSLHNSKDLSKDQHGRNCLLASYVHYVFRLPEVQRDVPKSGAPTALLDPRSYHTYGRTSAAAVSSKLLQARVMSSSNPDLAGTHSAADEEVKNIMSSKIADRNCSRMSYYCSGSSDAPSS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... DOCK8(81704)

General description

Dedicator of cytokinesis protein 8 or DOCK8 is a member of DOCK protein family that regulates cell development, adhesion, structure and movement. DOCK8 may also modulate the reorganization of actin filaments. Loss-of-function mutations in DOCK8 have been associated with combined immunodeficiency, whereas down-regulation of DOCK8 expression has been linked to pulmonary cancer . Anti-DOCK8 antibody is specific for DOCK8 in humans.

Immunogen

Dedicator of cytokinesis protein 8 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST79906.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fayhan J Alroqi et al.
Journal of clinical immunology, 37(8), 811-819 (2017-10-24)
The dedicator of cytokinesis 8 (DOCK8) deficiency is an autosomal recessive-combined immunodeficiency whose clinical spectra include recurrent infections, autoimmunity, malignancies, elevated serum IgE, eczema, and food allergies. Here, we report on patients with loss of function DOCK8 mutations with profound
Minnie Jacob et al.
Allergy, 74(2), 370-379 (2018-09-27)
Hyper-IgE syndromes (HIES) are a clinically overlapping, heterogeneous group of inborn errors of immunity characterized by elevated serum IgE level, eosinophilia, atopy, and immune dysregulation. Deficiency of DOCK8 protein is potentially a life-threatening autosomal recessive HIES and only curable with
Alicia S Wilson et al.
European journal of immunology, 49(5), 770-781 (2019-02-08)
Mutation of Dedicator of cytokinesis 8 (DOCK8) has previously been reported to provide resistance to the Th17 cell dependent EAE in mice. Contrary to expectation, we observed an elevation of Th17 cells in two different DOCK8 mutant mouse strains in
Jennifer R Yonkof et al.
Frontiers in immunology, 11, 884-884 (2020-07-07)
CARMIL2 deficiency is a rare combined immunodeficiency (CID) characterized by defective CD28-mediated T cell co-stimulation, altered cytoskeletal dynamics, and susceptibility to Epstein Barr Virus smooth muscle tumors (EBV-SMTs). Case reports associated with EBV-SMTs are limited. We describe herein a novel
Melanie Schauer et al.
Scientific reports, 8(1), 12332-12332 (2018-08-19)
The GTP-binding protein septin 7 is involved in various cellular processes, including cytoskeleton organization, migration and the regulation of cell shape. Septin 7 function in lymphocytes, however, is poorly characterized. Since the intracellular signaling role of septin 7 is dependent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service