Skip to Content
Merck
All Photos(6)

Documents

HPA002736

Sigma-Aldrich

Anti-CFAP57 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-FLJ32000, Anti-WDR65

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

PESNLVYWLWEKQKVMAIVRIDTQNNPVYQVSFSPQDNTQVCVTGNGMFKLLRFAEGTLKQTSFQRGEPQNYLAHTWVADDKIVVGTDTGKLFLFESGDQRWETSIMVKEPTNGSKSLDVIQESESLIEFPPVSSPLP

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... WDR65(149465)

General description

The heterotrimeric G protein WDR65 (WD repeat domain 65) is a member of the conservative WD repeat-containing family of proteins. It is present in all eukaryotes but absent in prokaryotes. It forms a circularized β propeller structure. The β subunit consists of two distinct regions: an amino-terminal α-helical region and WD repeat units which are arranged in a ring to form a propeller structure with seven blades. All blades are composed of antiparallel β sheet.

Immunogen

cilia and flagella associated protein 57

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

WDR65 (WD repeat domain 65) is associated with various biological functions such as signal transduction, transcription regulation, pre-mRNA splicing, cytoskeletal assembly, and vesicular traffic. It is also associated with different human diseases. The β subunit of the protein is responsible for transducing cellular response or signal across the plasma membrane.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86367

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E J Neer et al.
Nature, 371(6495), 297-300 (1994-09-22)
WD proteins are made up of highly conserved repeating units usually ending with Trp-Asp (WD). They are found in all eukaryotes but not in prokaryotes. They regulate cellular functions, such as cell division, cell-fate determination, gene transcription, transmembrane signalling, mRNA
T F Smith et al.
Trends in biochemical sciences, 24(5), 181-185 (1999-05-14)
Our knowledge of the large family of proteins that contain the WD repeat continues to accumulate. The WD-repeat proteins are found in all eukaryotes and are implicated in a wide variety of crucial functions. The solution of the three-dimensional structure
I Garcia-Higuera et al.
The Journal of biological chemistry, 273(15), 9041-9049 (1998-05-16)
The beta subunit of the heterotrimeric G proteins that transduce signals across the plasma membrane is made up of an amino-terminal alpha-helical segment followed by seven repeating units called WD (Trp-Asp) repeats that occur in about 140 different proteins. The
D Li et al.
Cellular and molecular life sciences : CMLS, 58(14), 2085-2097 (2002-01-30)
Defined by the presence of four or more repeating units containing a conserved core of approximately 40 amino acids that usually ending with tryptophan-aspartic acid (WD), WD-repeat proteins belong to a large and fast-expanding conservative protein family. As demonstrated by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service