Skip to Content
Merck
All Photos(2)

Documents

HPA001915

Sigma-Aldrich

Anti-TOP1MT antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-DNA topoisomerase I, mitochondrial precursor antibody produced in rabbit, Anti-TOP1mt antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

GYCILDGHQEKIGNFKIEPPGLFRGRGDHPKMGMLKRRITPEDVVINCSRDSKIPEPPAGHQWKEVRSDNTVTWLAAWTESVQNSIKYIMLNPCSKLKGETAWQKFETARRLRGFVDEIRSQYRADWKSREMKTRQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TOP1MT(116447)

Immunogen

DNA topoisomerase I, mitochondrial precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

TOP1MT (topoisomerase (DNA) I, mitochondrial) encodes a type IB topoisomerase. At the N-terminal end, it contains a mitochondrial localization sequence instead of a nuclear localization signal, thus, called as TOP1MT. In presence of divalent metal (Ca2+ or Mg2+) and alkaline pH, it shows optimum activity in several DNA metabolic processes. It is essentially required for DNA strand entanglements, chromatin compaction, and DNA decatenation during replication and recombination.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST85136

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stefan Sobek et al.
Experimental gerontology, 56, 135-141 (2014-01-21)
DNA topoisomerases pass DNA strands through each other, a function essential for all DNA metabolic processes that create supercoils or entanglements of DNA. Topoisomerases play an ambivalent role in nuclear genome maintenance: Deficiency compromises gene transcription, replication and chromosome segregation
Hongliang Zhang et al.
Biochimie, 89(4), 474-481 (2006-12-13)
Mitochondria are the only organelles containing metabolically active DNA besides nuclei. By analogy with the nuclear topoisomerases, mitochondrial topoisomerase activities are probably critical for maintaining the topology of mitochondrial DNA during replication, transcription, and repair. Mitochondrial diseases include a wide
H Zhang et al.
Proceedings of the National Academy of Sciences of the United States of America, 98(19), 10608-10613 (2001-08-30)
Tension generated in the circular mitochondrial genome during replication and transcription points to the need for mtDNA topoisomerase activity. Here we report a 601-aa polypeptide highly homologous to nuclear topoisomerase I. The N-terminal domain of this novel topoisomerase contains a
Thomas J Nicholls et al.
Molecular cell, 69(1), 9-23 (2018-01-02)
How mtDNA replication is terminated and the newly formed genomes are separated remain unknown. We here demonstrate that the mitochondrial isoform of topoisomerase 3α (Top3α) fulfills this function, acting independently of its nuclear role as a component of the Holliday junction-resolving
Marina Feric et al.
The EMBO journal, 40(6), e107165-e107165 (2021-02-24)
Mitochondria contain an autonomous and spatially segregated genome. The organizational unit of their genome is the nucleoid, which consists of mitochondrial DNA (mtDNA) and associated architectural proteins. Here, we show that phase separation is the primary physical mechanism for assembly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service