Skip to Content
Merck
All Photos(3)

Documents

HPA001400

Sigma-Aldrich

Anti-IMPDH2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-IMP dehydrogenase 2 antibody produced in rabbit, Anti-IMPD 2 antibody produced in rabbit, Anti-IMPDH-II antibody produced in rabbit, Anti-Inosine-5′-monophosphate dehydrogenase 2 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

RNAi knockdown
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

GFITDPVVLSPKDRVRDVFEAKARHGFCGIPITDTGRMGSRLVGIISSRDIDFLKEEEHDCFLEEIMTKREDLVVAPAGITLKEANEILQRSKKGKLPIVNEDDELVAIIARTDLKKNRDYPLASKD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IMPDH2(3615)

Immunogen

Inosine-5′-monophosphate dehydrogenase 2 recombinant protein epitope signature tag (PrEST)

Application

Anti-IMPDH2 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

IMPDH2 (inosine 5′-monophosphate dehydrogenase 2) encodes a rate-limiting enzyme involved in the de novo guanine nucleotide biosynthesis that catalyzes the NAD-dependent oxidation of inosine-5′-monophosphate (IMP) into xanthine-5′-monophosphate (XMP). It maintains the cellular guanine deoxy- and ribonucleotide pools that are required for the synthesis of DNA and RNA. It may be involved in some tumor progression and malignancy and may serve as a target for anticancer and immunosuppressive chemotherapy as it catalyzes an important step in purine nucleotide biosynthesis, which is necessary for the proliferation of lymphocytes.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST83044

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stefan J Barfeld et al.
Oncotarget, 6(14), 12587-12602 (2015-04-15)
The androgen receptor is a key transcription factor contributing to the development of all stages of prostate cancer (PCa). In addition, other transcription factors have been associated with poor prognosis in PCa, amongst which c-Myc (MYC) is a well-established oncogene
M D Sintchak et al.
Immunopharmacology, 47(2-3), 163-184 (2000-07-06)
The enzyme IMPDH is a homotetramer of approximately 55 kDa subunits and consists of a (beta/alpha)(8) barrel core domain and a smaller subdomain. The active site has binding pockets for the two substrates IMP and NAD. The enzymatic reaction of
S F Carr et al.
The Journal of biological chemistry, 268(36), 27286-27290 (1993-12-25)
Human IMP dehydrogenase, a target for anticancer and immunosuppressive chemotherapy, exists as two isoforms, types I and II. Nonfusion sequences of each isoform were overexpressed in an IMP dehydrogenase-deficient strain of Escherichia coli and purified to homogeneity. Both recombinant isoforms
Anna Bianchi-Smiraglia et al.
Nature methods, 14(10), 1003-1009 (2017-09-05)
GTP is a major regulator of multiple cellular processes, but tools for quantitative evaluation of GTP levels in live cells have not been available. We report the development and characterization of genetically encoded GTP sensors, which we constructed by inserting
Joseph A Wawrzyniak et al.
Cell reports, 5(2), 493-507 (2013-10-22)
Melanoma is one of the most aggressive types of human cancers, and the mechanisms underlying melanoma invasive phenotype are not completely understood. Here, we report that expression of guanosine monophosphate reductase (GMPR), an enzyme involved in de novo biosynthesis of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service