Skip to Content
Merck
All Photos(6)

Documents

HPA000614

Sigma-Aldrich

ANTI-CDC45 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CDC45-related protein, Anti-CDC45L, Anti-Cdc45, Anti-PORC-PI-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

MFVSDFRKEFYEVVQSQRVLLFVASDVDALCACKILQALFQCDHVQYTLVPVSGWQELETAFLEHKEQFHYFILINCGANVDLLDILQPDEDTIFFVCDTHRPVNVVNVYNDTQIKLLIKQDDDLEVPAYEDIFRDEEEDEEHSGNDSDGSEPSEKRTRL

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDC45L(8318)

Immunogen

CDC45-related protein recombinant protein epitope signature tag (PrEST)

Application

Anti-CDC45L antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

CDC45 (cell division cycle 45) is an essential protein in the initiation of chromosomal DNA replication. It is a component of the multiprotein complex consisting of Cdc6/Cdc18, the minichromosome maintenance proteins (MCMs) and DNA polymerase. It interacts with MCM7 and DNA polymerase α and assists in the loading of DNA polymerase α onto chromatin. It is ubiquitylated and degraded during apoptosis. Deletion of CD45 gene causes DiGeorge syndrome with parathyroid hypoplasia, thymic aplasia, or hypoplasia and congenital cardiac abnormalities.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73431

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junichiro Takaya et al.
Journal of biochemistry, 153(4), 381-388 (2013-02-01)
CDC45, which plays a role in eukaryotic DNA replication, is a member of the CMG (CDC45/MCM2-7/GINS) complex that is thought to function as a replicative DNA helicase. However, the biochemical properties of CDC45 are not fully understood. We systematically examined
Anna Szambowska et al.
Nucleic acids research, 42(4), 2308-2319 (2013-12-03)
The cell division cycle protein 45 (Cdc45) represents an essential replication factor that, together with the Mcm2-7 complex and the four subunits of GINS, forms the replicative DNA helicase in eukaryotes. Recombinant human Cdc45 (hCdc45) was structurally characterized and its
S Mimura et al.
The EMBO journal, 17(19), 5699-5707 (1998-10-02)
At the onset of S phase, chromosomal replication is initiated by the loading of DNA polymerase alpha onto replication origins. However, the molecular mechanisms for controlling the initiation are poorly understood. Using Xenopus egg extract, we report here the identification
Ziyan Chen et al.
Digestive surgery, 39(1), 24-31 (2021-11-18)
The aim of this study is selecting the hub genes associated with hepatocellular carcinoma (HCC) to construct a Cox regression model for predicting prognosis in HCC patients. Using HCC patient data from the ICGC and TCGA databases, screened for 40
M Lei et al.
Journal of cell science, 114(Pt 8), 1447-1454 (2001-04-03)
The exact duplication of a genome once per cell division is required of every proliferating cell. To achieve this goal, eukaryotes adopt a strategy that limits every replication origin to a single initiation event within a narrow window of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service