Skip to Content
Merck
All Photos(4)

Key Documents

HPA000600

Sigma-Aldrich

Anti-ABHD4 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-α/β-Hydrolase 4 antibody produced in rabbit, Anti-lyso-N-Acylphosphatidylethanolamine lipase antibody produced in rabbit, Anti-Abhydrolase domain-containing protein 4 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Estimated to ship on18 April 2025



Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

$1,040.00


Estimated to ship on18 April 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

ADDLEQQSQGWLSSWLPTWRPTSMSQLKNVEARILQCLQNKFLARYVSLPNQNKIWTVTVSPEQNDRTPLVMVHGFGGGVGLWILNMDSLSARRTLHTFDLLGFGRSSRPAFPRDPEGAEDEFVTSIETWRETMGIPSMILLGHSLGGF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ABHD4(63874)

Immunogen

Abhydrolase domain-containing protein 4 recombinant protein epitope signature tag (PrEST)

Application

Anti-ABHD4 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

ABHD4 (αβ hydrolase domain containing 4) is a genetic regulator of anoikis resistance and sensitivity. It acts as a (lyso)N-acyl phosphatidylethanolamines (NAPE) -selective lipase in the NAPE-PLD (phospholipase D)-independent pathway for NAE (N-acyl ethanolamines) biosynthesis. NAEs are signaling lipids that function in various physiological processes such as, nociception, cognition, anxiety, appetite, and inflammation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70348

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Short time incubation at low temperature for retrieval of some antigens from formalin-fixed and paraffin-embedded tissues.
Abdulai, S M, et al.
Journal of Histotechnology, 35, 74-79 (2012)
Craig D Simpson et al.
Apoptosis : an international journal on programmed cell death, 17(7), 666-678 (2012-04-11)
Acquisition of resistance to anchorage dependant cell death, a process termed anoikis, is a requirement for cancer cell metastasis. However, the molecular determinants of anoikis resistance and sensitivity are poorly understood. To better understand resistance to anoikis we conducted a
Gabriel M Simon et al.
The Journal of biological chemistry, 281(36), 26465-26472 (2006-07-05)
N-Acyl ethanolamines (NAEs) are a large class of signaling lipids implicated in diverse physiological processes, including nociception, cognition, anxiety, appetite, and inflammation. It has been proposed that NAEs are biosynthesized from their corresponding N-acyl phosphatidylethanolamines (NAPEs) in a single enzymatic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service