Skip to Content
Merck
All Photos(1)

Key Documents

H9902

Sigma-Aldrich

Heparan sulfate fast-moving fraction sodium salt from porcine intestinal mucosa

≥90% (electrophoresis)

Synonym(s):

Heparitin sulfate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$56.10

Vendor SKU: PC9251-1G

$56.10


Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
1 G
$56.10

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201

Vendor SKU: PC9251-1G

$56.10


Ships in 7 days - from Aldrich Partner.

Assay

≥90% (electrophoresis)

storage temp.

2-8°C

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paul Proost et al.
The Journal of experimental medicine, 205(9), 2085-2097 (2008-08-20)
Biological functions of proteins are influenced by posttranslational modifications such as on/off switching by phosphorylation and modulation by glycosylation. Proteolytic processing regulates cytokine and chemokine activities. In this study, we report that natural posttranslational citrullination or deimination alters the biological
U Benelli et al.
Experimental eye research, 67(2), 133-142 (1998-09-12)
The purpose of this study was to evaluate the inhibitory activity of the heparan sulfate suleparoide on vascular cell growth in vitro and angiogenesis in vivo. Human HUV-EC-C endothelial cell proliferation and microvessel sprouting from cultured rat aortic rings were
Comparative pharmacologic profile of a glycosaminoglycan mixture, Sulodexide, and a chemically modified heparin derivative, Suleparoide.
D D Callas et al.
Seminars in thrombosis and hemostasis, 19 Suppl 1, 49-57 (1993-01-01)
A Lepri et al.
Ophthalmic research, 28(3), 176-183 (1996-01-01)
Intra-ocular fibrin deposition following ocular surgery is a serious postoperative complication. The current management of severe postoperative fibrin response is usually ineffective. A quantitative model of fibrin deposition in the rabbit anterior chamber was used to assess the efficacy of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service