Skip to Content
Merck
All Photos(1)

Key Documents

H9897

Sigma-Aldrich

HEPES hemisodium salt

dry powder

Synonym(s):

4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid hemisodium salt, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid) hemisodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

10 PKG
$295.00

$295.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 PKG
$295.00

About This Item

Empirical Formula (Hill Notation):
C8H17.5N2O4S· 0.5Na
CAS Number:
Molecular Weight:
249.30
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

$295.00


Check Cart for Availability

Request a Bulk Order

form

dry powder

pH

7.0-8.0

useful pH range

6.8-8.2

pKa 

7.5

storage temp.

room temp

SMILES string

[Na+].OCCN1CCN(CC1)CCS([O-])(=O)=O

InChI

1S/C8H18N2O4S.Na/c11-7-5-9-1-3-10(4-2-9)6-8-15(12,13)14;/h11H,1-8H2,(H,12,13,14);/q;+1/p-1

InChI key

RDZTWEVXRGYCFV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Packaging

foil pouches

Reconstitution

Contents of each pouch dissolved in 1 liter deionized water will yield 0.1 M sodium HEPES buffer, pH 7.5 at 25 °C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mario Vieweger et al.
Biophysical journal, 114(8), 1762-1775 (2018-04-26)
Structural RNA domains are widely involved in the regulation of biological functions, such as gene expression, gene modification, and gene repair. Activity of these dynamic regions depends sensitively on the global fold of the RNA, in particular, on the binding
Benjamin S Waldman et al.
Cell, 180(2), 359-372 (2020-01-21)
Toxoplasma gondii chronically infects a quarter of the world's population, and its recrudescence can cause life-threatening disease in immunocompromised individuals and recurrent ocular lesions in the immunocompetent. Acute-stage tachyzoites differentiate into chronic-stage bradyzoites, which form intracellular cysts resistant to immune

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service