Skip to Content
Merck
All Photos(5)

Key Documents

H9411

Sigma-Aldrich

Anti-Histone Deacetylase 4 (HDAC4) (ML-19) antibody produced in rabbit

enhanced validation

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-AHO3, Anti-BDMR, Anti-HA6116, Anti-HD4, Anti-HDAC-4, Anti-HDAC-A, Anti-HDACA

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$683.00

$683.00


Estimated to ship on01 April 2025



Select a Size

Change View
200 μL
$683.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$683.00


Estimated to ship on01 April 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~140 kDa (additional weaker bands may be present)

species reactivity

rat, human, mouse

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoprecipitation (IP): 10-20 μg using RIPA extract of HeLa nuclei
indirect immunofluorescence: 1:250 using HeLa cells
microarray: suitable
western blot: 1:1,000 using whole extracts of mouse NIH3T3 cells
western blot: 1:500 using whole extracts of rat brain

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HDAC4(9759)
mouse ... Hdac4(208727)
rat ... Hdac4(363287)

General description

Mammalian histone deacetylases (HDACs) can be divided into three classes according to sequence homology. Class II consists of the yeast Hda1-like proteins histone deacetylase 4,5,6,7,8,9, 10 (HDAC4, HDAC5, HDAC6, HDAC7, HDAC9, and HDAC10.

Immunogen

synthetic peptide corresponding to amino acid residues 1-19 of human HDAC4 with C-terminal added lysine conjugated to KLH.

Application

Anti-Histone Deacetylase 4 (HDAC4) (ML-19) antibody produced in rabbit has been used in:
  • immunoblotting
  • immunoprecipitation
  • immunofluorescence
  • immunostaining
  • immunohistochemistry
  • chromatin immunoprecipitation

Biochem/physiol Actions

Histone deacetylase 4 (HDAC4) exhibits intrinsic nuclear import and export signals for its dynamic nucleocytoplasmic shuttling. Its association with 14-3-3 and monocyte enhancer factor 2 (MEF2) proteins can disturb shuttling and thus directs HDAC4 to the cytoplasm and the nucleus, respectively.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enzymatic activity associated with class II HDACs is dependent on a multiprotein complex containing HDAC3 and SMRT/N-CoR
Fischle W, et al.
Molecular Cell, 9(1), 45-57 (2002)
Lilian Varricchio et al.
The international journal of biochemistry & cell biology, 50, 112-122 (2014-03-07)
To clarify the role of HDACs in erythropoiesis, expression, activity and function of class I (HDAC1, HDAC2, HDAC3) and class IIa (HDAC4, HDAC5) HDACs during in vitro maturation of human erythroblasts were compared. During erythroid maturation, expression of HDAC1, HDAC2
Stress-induced epigenetic transcriptional memory of acetylcholinesterase by HDAC4
Sailaja BS, et al.
Proceedings of the National Academy of Sciences of the USA, 109(52), E3687-E3695 (2012)
Histone deacetylase inhibitors preserve white matter structure and function during ischemia by conserving ATP and reducing excitotoxicity
Baltan S, et al.
The Journal of Neuroscience, 31(11), 3990-3999 (2011)
A H Wang et al.
Molecular and cellular biology, 21(17), 5992-6005 (2001-08-04)
Nucleocytoplasmic trafficking of histone deacetylase 4 (HDAC4) plays an important role in regulating its function, and binding of 14-3-3 proteins is necessary for its cytoplasmic retention. Here, we report the identification of nuclear import and export sequences of HDAC4. While

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service