Skip to Content
Merck
All Photos(1)

Key Documents

H9383

Sigma-Aldrich

Anti-Horse Serum antibody produced in rabbit

fractionated antiserum, lyophilized powder

Synonym(s):

Anti-Serum, horse

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
$176.00

$176.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 VIAL
$176.00

About This Item

MDL number:
UNSPSC Code:
12352203

$176.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

fractionated antiserum

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

horse

packaging

vial of 2 mL lyophilized antiserum

technique(s)

immunoelectrophoresis: suitable

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Horse serum antibody is purified from pooled serum from normal donor herd.

Physical form

Lyophilized from 0.01 M phosphate buffered saline, pH 7.2

Reconstitution

Reconstitute with 2 ml deionized water.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service