All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C8H12N4O3
CAS Number:
Molecular Weight:
212.21
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Recommended Products
storage temp.
−20°C
SMILES string
NC(Cc1cnc[nH]1)C(=O)NCC(O)=O
Biochem/physiol Actions
Histidylleucine (His-Leu); histidylglycine (His-Gly) and histidylserine (His-Ser) are N-terminal imidazole containing dipeptides used to study binding of metals such as copper, nickel and zinc.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Anoja P Wickrama Arachchilage et al.
The Journal of chemical physics, 136(12), 124301-124301 (2012-04-03)
We have investigated the electronic structure of three cyclic dipeptides: cyclo(Histidyl-Glycyl) (cHisGly), cyclo(Tyrosyl-Prolyl) (cTyrPro), and cyclo(Phenylalanyl-Phenylalanyl) (cPhePhe) in the vapor phase, by means of photoemission spectroscopy and theoretical modeling. The last compound was evaporated from the solid linear dipeptide, but
Brandon I Macdonald et al.
Rapid communications in mass spectrometry : RCM, 22(18), 2946-2954 (2008-08-30)
Pathways for proton transfer in the histidylglycine cation are examined in the gas-phase environment with the goal of understanding the mechanism by which protons may become mobile in proteins with basic amino acid residues. An extensive search of the potential
Terézia Szabó-Plánka et al.
Inorganic chemistry, 41(13), 3483-3490 (2002-06-25)
Twelve ESR-active (and one inactive) copper(II) complexes of L-histidylglycine (HL) were characterized via their formation (micro)constants and ESR parameters obtained by two-dimensional ESR spectroscopic evaluation in aqueous solution. In strongly acidic media, the ligand is coordinated through its N-terminal donor
Brandon J Green et al.
Inorganic chemistry, 43(4), 1463-1471 (2004-02-18)
Self-decomposition of the nickel(III) doubly deprotonated peptide complex of Gly2HisGly occurs by base-assisted oxidation of the peptide. At < or =p[H+] 7.0, the major pathway is a four-electron oxidation (via 4 Ni(III) complexes) at the alpha carbon of the N-terminal
G Kilian et al.
Die Pharmazie, 68(3), 207-211 (2013-04-06)
Cyclic dipeptides have been well characterized for their biological activity, including antimicrobial and anticancer activities. Cyclo(His-Gly) and cyclo(His-Ala) have also recently demonstrated significant anticancer activity against a range of cell lines, however, as a result of their physicochemistry, namely high
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service