Skip to Content
Merck
All Photos(1)

Key Documents

H8631

Sigma-Aldrich

5-Hydroxymethyl-2′-deoxyuridine

≥95% (HPLC)

Synonym(s):

5-HMdU, HMdUdr

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2O6
CAS Number:
Molecular Weight:
258.23
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

Assay

≥95% (HPLC)

form

powder

solubility

water: 50 mg/mL, clear, colorless to very faintly yellow

storage temp.

−20°C

SMILES string

OCC1OC(CC1O)N2C=C(CO)C(=O)NC2=O

InChI

1S/C10H14N2O6/c13-3-5-2-12(10(17)11-9(5)16)8-1-6(15)7(4-14)18-8/h2,6-8,13-15H,1,3-4H2,(H,11,16,17)

InChI key

IPAVKOYJGUMINP-UHFFFAOYSA-N

Application

5-Hydroxymethyl-2′-deoxyuridine (HMdUdr) is a used as a marker for oxidative nucleic acid (DNA) damage. HMdUdr may be used as an antigen to create anti-HMdUdr antibodies.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Mitchell Harman et al.
Free radical biology & medicine, 35(10), 1301-1309 (2003-11-11)
To assess novel liquid chromatography/mass spectrometric methods for measuring oxidative damage to nucleic acids and lipids, we compared urinary excretion of 8-hydroxy-2'-deoxyguanosine (8-OHdG), 5-hydroxymethyl-2'-deoxyuridine (5-OHmU), and 8-hydroxyguanosine (8-OxoG), and an isoprostane, 8-iso-prostaglandin F(2)alpha (IsopF(2)alpha) in 234 healthy men (n =
Jin Wang et al.
Analytical chemistry, 83(6), 2201-2209 (2011-02-18)
The purpose of our study was to develop suitable methods to quantify oxidative DNA lesions in the setting of transition metal-related diseases. Transition metal-driven Fenton reactions constitute an important endogenous source of reactive oxygen species (ROS). In genetic diseases with
I Kato et al.
Biomarkers : biochemical indicators of exposure, response, and susceptibility to chemicals, 11(2), 143-152 (2006-06-13)
Oxidative stress has been implicated in the pathogenesis of various chronic diseases, such as cancer, cardiovascular disease and inflammatory conditions, as well as in ageing. Although a number of markers are now available, little is known about the reliability of
Joshy Joseph et al.
Chemical communications (Cambridge, England), 46(42), 7872-7878 (2010-09-11)
The feature article is a review of the reaction of thymine in the one-electron oxidation of duplex DNA. Oxidation of DNA causes chemical reactions that result in remote damage (mutation) to a nucleobase. Normally this reaction occurs at guanine, but
Kiran S Toti et al.
Bioorganic & medicinal chemistry, 21(1), 257-268 (2012-12-04)
We report the synthesis of 5'-modified thymidines (16, 18, 21, 23) and 5,5'-bis-substituted 2'-deoxyuridine analogues (30, 47) as inhibitors of thymidine monophosphate kinase of Mycobacterium tuberculosis (TMPKmt). These analogues were evaluated for their capacity to inhibit TMPKmt and solely two

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service