Skip to Content
Merck
All Photos(1)

Documents

H7789

Sigma-Aldrich

Anti-acetyl-Histone H1.4 (Ac-Lys26) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-H1 Histone family, member 4, Anti-H1.4K26ac, Anti-H1K26ac, Anti-H1e, Anti-Histone 1, Anti-Histone H1b

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~35 kDa

species reactivity

human

technique(s)

western blot: 0.5-1 μg/mL using acid-extracted fraction of HL60 cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

acetylation (Lys26)

Gene Information

human ... HIST1H1E(3008)

General description

In mammalian cells, four histone H1 variants (H1.2 to H1.5) are present in all somatic cells, and a fifth (H1.1) is restricted to thymus, testis, and spleen and possibly lymphocytic and neuronal cells. Histone H1.4 is di-methylated or acetylated at Lys26. Lys26 is located within the flexible N-terminal domain of H1.4, just preceding the globular domain.

Immunogen

synthetic acetylated peptide corresponding to amino acids 22-33 (Ac-Lys26) of human histone H1.4.

Application

Anti-acetyl-Histone H1.4 (Ac-Lys26) antibody produced in rabbit is suitable for western blot at a concentration of 0.5-1μg/mL using acid-extracted fraction of HL60 cells.
Anti-acetyl-Histone H1.4 (Ac-Lys26) antibody produced in rabbit has been used in western blotting.

Biochem/physiol Actions

H1.4 at lysine residue 26 represses transcription. Linker histone H1 binds to DNA between nucleosomal core particles and is involved in establishing and maintaining higher order chromatin structures. Histone modifications are thought to play an important role in cancer and disease.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dynamic histone H1 isotype 4 methylation and demethylation by histone lysine methyltransferase G9a/KMT1C and the Jumonji domain-containing JMJD2/KDM4 proteins
Trojer P, et al.
The Journal of Biological Chemistry, 284(13), 8395-8405 (2009)
Mechanistic and structural studies of KDM-catalysed demethylation of histone 1 isotype 4 at lysine 26
Walport LJ, et al.
Febs Letters, 592(19), 3264-3273 (2018)
Anti-correlation between longevity gene SirT1 and Notch signaling in ascending aorta biopsies from patients with bicuspid aortic valve disease
Sciacca S, et al.
Heart and Vessels, 28(2), 268-275 (2013)
S Khochbin
Gene, 271(1), 1-12 (2001-06-19)
Genes encoding linker histone variants have evolved to link their expression to signals controlling the proliferative capacities of cells, i.e. cycling and growth-arrested cells express distinct and specific H1 subtypes. In metazoan, these variants show a tripartite structure, with considerably
Jerome Bonnefont et al.
Neuron, 103(6), 1096-1108 (2019-07-30)
During neurogenesis, progenitors switch from self-renewal to differentiation through the interplay of intrinsic and extrinsic cues, but how these are integrated remains poorly understood. Here, we combine whole-genome transcriptional and epigenetic analyses with in vivo functional studies to demonstrate that Bcl6

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service