Skip to Content
Merck
All Photos(1)

Documents

H7289

Sigma-Aldrich

Hydroxyphenyl fluorescein

Synonym(s):

2-[6-(4′-hydroxy)phenoxy-3H-xanthen-3-on-9-yl]benzoic acid, HPF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H16O6
Molecular Weight:
424.40
UNSPSC Code:
12352200

storage condition

protect from light

fluorescence

λex 490 nm; λem 515 nm

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Fluorescent reagent for the detection of highly reactive oxygen species (hROS). Fluorescence intensity increases after the immediate reaction with hROS such as hydroxyl radical and peroxynitrite. Since HPF does not react with nitric oxide, superoxide and hydrogen peroxide, it can differentially detect peroxynitrite. HPF is resistant to light-induced autoxidation.

comparable product

Product No.
Description
Pricing

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service