Skip to Content
Merck
All Photos(1)

Key Documents

H7160

Sigma-Aldrich

Hepatitis C virus core antigen−Fluorescein

~1 mg/mL, ≥95% (SDS-PAGE), recombinant, expressed in E. coli

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

recombinant

expressed in E. coli

Quality Level

Assay

≥95% (SDS-PAGE)

mol wt

22 kDa (plus 114 kDa β-Gal tag)

concentration

~1 mg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

hepatitis C virus ... HCVgp1(951475)

Biochem/physiol Actions

Positive control for HCV antibodies.

Other Notes

HCV core antigen [2-192]-Galactosidase-tagged, fluorescein conjugate.

Physical form

Solution in 1.2 M Urea, 20 mM Tris-HCl, pH 8.0, 10 mM 2-mercaptoethanol.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inés Romero-Brey et al.
mBio, 6(4), e00759-e00759 (2015-07-15)
Induction of membrane rearrangements in the cytoplasm of infected cells is a hallmark of positive-strand RNA viruses. These altered membranes serve as scaffolds for the assembly of viral replication factories (RFs). We have recently shown that hepatitis C virus (HCV)
Margaret A Scull et al.
PLoS pathogens, 11(11), e1005297-e1005297 (2015-11-21)
The hepatitis C virus (HCV) p7 protein is required for infectious virus production via its role in assembly and ion channel activity. Although NMR structures of p7 have been reported, the location of secondary structural elements and orientation of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service