Skip to Content
Merck
All Photos(1)

Key Documents

H6042

Sigma-Aldrich

n-Heptanoyl coenzyme A lithium salt

≥95% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$597.00

$597.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$597.00

About This Item

UNSPSC Code:
41106305
NACRES:
NA.51

$597.00


Estimated to ship on29 May 2025


Request a Bulk Order

Assay

≥95% (HPLC)

storage temp.

−20°C

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service