Skip to Content
Merck
All Photos(1)

Key Documents

H6039

Sigma-Aldrich

1-(4-Hexyphenyl)-2-propane-1-one

≥98% (HPLC), oil

Synonym(s):

L3 Antagonist TR-SIR-2, cpd L3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H20O
CAS Number:
Molecular Weight:
216.32
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

oil

solubility

DMSO: >5 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

CCCCCCc1ccc(cc1)C(=O)C=C

InChI

1S/C15H20O/c1-3-5-6-7-8-13-9-11-14(12-10-13)15(16)4-2/h4,9-12H,2-3,5-8H2,1H3

InChI key

IINHTEWASPUCMH-UHFFFAOYSA-N

Biochem/physiol Actions

Small molecular irreversible covalent inhibitor of thyroid hormone receptor-β (TRβ) interaction with coactivator (SRC2).

Features and Benefits

This compound is featured on the Nuclear Receptors (Non-Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service