Skip to Content
Merck
All Photos(1)

Key Documents

H5520

Sigma-Aldrich

17α-Hydroxyprogesterone hexanoate

≥98%

Synonym(s):

17α-Hydroxyprogesterone caproate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H40O4
CAS Number:
Molecular Weight:
428.60
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98%

SMILES string

[H][C@@]12CCC3=CC(=O)CC[C@]3(C)[C@@]1([H])CC[C@@]4(C)[C@@]2([H])CC[C@]4(OC(=O)CCCCC)C(C)=O

InChI

1S/C27H40O4/c1-5-6-7-8-24(30)31-27(18(2)28)16-13-23-21-10-9-19-17-20(29)11-14-25(19,3)22(21)12-15-26(23,27)4/h17,21-23H,5-16H2,1-4H3/t21-,22+,23+,25+,26+,27+/m1/s1

InChI key

DOMWKUIIPQCAJU-LJHIYBGHSA-N

Gene Information

human ... PGR(5241)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Shows significant reduction of pre-term delivery by mothers with a history of premature delivery, with no detectable harm to the infant.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shringi Sharma et al.
Drug metabolism and disposition: the biological fate of chemicals, 41(2), 296-304 (2012-11-07)
Little information is available in the literature regarding the expression and activity of transporters in fetal human liver or cultured cells. A synthetic progesterone structural analog, 17α-hydroxyprogesterone caproate (17-OHPC), is used in the prevention of spontaneous abortion in women with
Faris Mujezinovic et al.
The Cochrane database of systematic reviews, 8(8), CD008678-CD008678 (2012-08-17)
Currently, the techniques for amniocentesis and chorionic villus sampling (CVS) tend to be described in local and national guidelines, but certain aspects, including the choice of instruments, is predominantly based upon the operator's personal preference. A survey of practice in
Mary Hines et al.
Clinical drug investigation, 33(3), 223-227 (2013-02-16)
Intramuscular 17 α-hydroxyprogesterone caproate (Makena(®)), a synthetic progestin, is indicated to reduce the risk of preterm birth in women with a singleton pregnancy who have a history of a singleton spontaneous preterm birth. Makena(®) reduces the risk of preterm birth
John M O'Brien
American journal of perinatology, 29(9), 665-672 (2012-07-10)
The safety of supplemental progestin therapy during pregnancy reflects whether an agent exclusively promotes or potentially inhibits progestational cellular functions and whether treatment incites a metabolic derangement or other pathophysiology to initiate rare untoward events. No safety signal has been
Jeff M Szychowski et al.
The journal of maternal-fetal & neonatal medicine : the official journal of the European Association of Perinatal Medicine, the Federation of Asia and Oceania Perinatal Societies, the International Society of Perinatal Obstetricians, 25(12), 2686-2689 (2012-08-15)
To assess cerclage benefit in women with short cervix also receiving 17-α-hydroxyprogesterone caproate (17P) to prevent recurrent preterm birth (PTB). Secondary analysis of a multicenter trial of ultrasound-indicated cerclage for shortened cervical length (CL). Women with prior spontaneous PTB at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service