H4409
EX-CELL® CD Hybridoma Medium, Chemically Defined
Animal-component free, with sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for hybridoma, suitable for cell culture
Synonym(s):
Hybridoma Medium
Select a Size
$255.00
Estimated to ship on01 June 2025
Select a Size
About This Item
$255.00
Estimated to ship on01 June 2025
Recommended Products
description
for research or for further manufacturing use
sterility
sterile-filtered
form
liquid
technique(s)
cell culture | hybridoma: suitable
cell culture | mammalian: suitable
impurities
endotoxin, tested
components
glucose: 2.84 g/L
NaHCO3: yes
L-glutamine: no
shipped in
ambient
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Features and Benefits
Developed to meet the needs of biotechnology manufacturing, this medium supports rapid initial cell growth and high levels of antibody expression. It also supports high cell densities at high viability for extended periods without the use of animal-derived proteins.
Caution
Reconstitution
Legal Information
related product
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service