Skip to Content
Merck
All Photos(1)

Key Documents

H3541

Sigma-Aldrich

HIPK3 (163-562), active, His tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

DYRK6, FIST3, PKY, YAK1

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$537.00

$537.00


Estimated to ship on27 April 2025


Request a Bulk Order

Select a Size

Change View
10 μG
$537.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$537.00


Estimated to ship on27 April 2025


Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

41-56 nmol/min·mg

mol wt

~49 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... HIPK3(10114)

General description

HIPK3 (homeodomain-interacting protein kinase 3) is strongly expressed in the heart and muscle tissues. It is up-regulated in multidrug-resistant cells.[1]

Biochem/physiol Actions

HIPK3 (homeodomain-interacting protein kinase 3) is a nuclear serine/threonine protein kinase and mainly controls death receptor signaling. HIPK3 is involved in the transcriptional corepression, cell differentiation, growth suppression and apoptosis.[2][3] It is up-regulated in prostate cancer cells, resulting in increased resistance to Fas receptor-mediated apoptosis by disturbing association between FADD (fas-associated death domain) and caspase 8.[2]

Physical form

Supplied in 50 mM Sodium Phosphate, pH 7.0, with 150 mM imidazole, 0.1 mM PMSF, 0.2 mM DTT, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D A Begley et al.
Gene, 200(1-2), 35-43 (1997-12-31)
We have previously shown that several protein kinases are present in higher activity levels in multidrug resistant cell lines, such as KB-V1. We have now isolated a gene that codes for a putative protein kinase, PKY, of over 130 kDa
Hsin-Chieh Lan et al.
Molecular and cellular biology, 27(6), 2027-2036 (2007-01-11)
Steroids are synthesized in adrenal glands and gonads under the control of pituitary peptides. These peptides bind to cell surface receptors to activate the cyclic AMP (cAMP) signaling pathway leading to an increase of steroidogenic gene expression. Exactly how cAMP
James F Curtin et al.
The Journal of biological chemistry, 279(17), 17090-17100 (2004-02-10)
Elevated endogenous JNK activity and resistance to Fas receptor-mediated apoptosis have recently been implicated in progression of prostate cancer and can promote resistance to apoptosis in response to chemotherapeutic drugs. In addition, JNK has been demonstrated to promote transformation of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service