Skip to Content
Merck
All Photos(1)

Key Documents

H2153

Sigma-Aldrich

HABA/Avidin Reagent

lyophilized powder

Synonym(s):

HABA Reagent

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
$101.00

$101.00


Available to ship on02 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 VIAL
$101.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

$101.00


Available to ship on02 April 2025Details


Request a Bulk Order

form

lyophilized powder

Quality Level

usage

sufficient for ≤10 assays

color

orange

suitability

suitable for spectrophotometric determination of biotin and biotin conjugates in a wide variety of samples

storage temp.

2-8°C

General description

4-Hydroxyazobenzene-2-carboxylic acid (HABA) is an avidin-binding dye that can be stoichiometrically displaced by biotin. The subsequent change in absorption at A500 provides a basis for spectrophotometric determination of biotin levels in biological samples.
HABA/Avidin Reagent is specific for chromogenic applications that depend upon quantification of biotin.

Application

HABA/Avidin Reagent is suitable for spectrophotometric determination of biotin and biotin conjugates in a wide variety of samples.
Reagent is capable of detecting biotin at μMolar concentrations (i.e., 1 nmole per ml). Typically used to quantitate biotin in biotinylated compounds.

Reconstitution

Prepares a ready-to-use HABA/avidin solution in buffered saline, pH approx. 7.3, with preservative when reconstituted with 10 ml deionized water.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

related product

Product No.
Description
Pricing

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Einspanier et al.
Biology of reproduction, 61(2), 512-520 (1999-07-20)
Relaxin is a peptide hormone with a broad range of biological activities, related not only to parturition and lactation but possibly also to decidualization, implantation, and early pregnancy. The present study was designed to investigate the secretion pattern of relaxin
R S Niedbala et al.
Journal of biochemical and biophysical methods, 13(4-5), 205-210 (1986-11-01)
Parameters and conditions of an enzyme based assay for biotin and avidin are presented. Biotinylated glucose-6-phosphate dehydrogenase when complexed with avidin becomes inactivated. Thus it was possible to construct a competitive assay system for biotin. The assay is sensitive between

Questions

1–2 of 2 Questions  
  1. I would like to know how many assays can be done with a VIAL KIT and how much material it contains,

    1 answer
    1. This product is sufficient for ≤10 assays. When reconstituted with 10 ml deionized water, each vial will yield a solution with a pH of approximately 7.3, containing the following components: 0.3 mM 4-Hydroxyazobenzene-2-carboxylic acid (HABA), 0.45 mg/ml avidin, 0.3 M sodium chloride (NaCl), 0.01 M N-[2-Hydroxyethyl]piperazine-NN-[2-ethanesulfonic acid] (HEPES), a buffer with a pKa of 7.5, 0.01 M magnesium chloride (MgCl2), and 0.02% sodium azide as a preservative.

      Helpful?

  2. how much HABA is in the vial

    1 answer
    1. When reconstituted to 10mL, the vial will contain 0.3 mM HABA. Based on the molecular weight of HABA, 242.23 gmol, the calculated mass per vial is 726.69 ug. Please see the link below to review additional informaition available in the product datasheet:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/378/451/h2153dat.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service