Skip to Content
Merck
All Photos(1)

Documents

H2003

Sigma-Aldrich

Hydrindantin hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H10O6
CAS Number:
Molecular Weight:
322.27
Beilstein:
1895666
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

SMILES string

O.O.OC1(C(=O)c2ccccc2C1=O)C3(O)C(=O)c4ccccc4C3=O

InChI

1S/C18H10O6.2H2O/c19-13-9-5-1-2-6-10(9)14(20)17(13,23)18(24)15(21)11-7-3-4-8-12(11)16(18)22;;/h1-8,23-24H;2*1H2

InChI key

QIAIXVRAKYAOGJ-UHFFFAOYSA-N

Application

For photometric determination of amino acids and similar compounds.

Linkage

Reduced form of ninhydrin.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amino acid analysis: aqueous dimethyl sulfoxide as solvent for the ninhydrin reaction.
S Moore
The Journal of biological chemistry, 243(23), 6281-6283 (1968-12-10)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service