Skip to Content
Merck
All Photos(1)

Key Documents

H1542

Sigma-Aldrich

HPI-1 hydrate

Synonym(s):

2-Methoxyethyl 1,4,5,6,7,8-hexahydro-4-(3-hydroxyphenyl)-7-(2-methoxyphenyl)-2-methyl-5-oxo-3-quinolinecarboxylate, Hedgehog Pathway Inhibitor 1

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$136.00
25 MG
$554.00

$136.00


Check Cart for Availability
New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
5 MG
$136.00
25 MG
$554.00

About This Item

Empirical Formula (Hill Notation):
C27H29NO6 · xH2O
CAS Number:
Molecular Weight:
463.52 (anhydrous basis)
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

$136.00


Check Cart for Availability
New, lower price on this item!

Request a Bulk Order

storage condition

desiccated

storage temp.

2-8°C

SMILES string

O.COCCOC(=O)C1=C(C)NC2=C(C1c3cccc(O)c3)C(=O)CC(C2)c4ccccc4OC

InChI

1S/C27H29NO6.H2O/c1-16-24(27(31)34-12-11-32-2)25(17-7-6-8-19(29)13-17)26-21(28-16)14-18(15-22(26)30)20-9-4-5-10-23(20)33-3;/h4-10,13,18,25,28-29H,11-12,14-15H2,1-3H3;1H2

InChI key

BOASXIAGFOONSK-UHFFFAOYSA-N

Biochem/physiol Actions

HPI-1 is a Hedgehog pathway inhibitor; Gli1/Gli2 inhibitor.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service