Skip to Content
Merck
All Photos(1)

Documents

GW22927F

Sigma-Aldrich

Anti-α-1-Acid Glycoprotein 1 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-AGP1, Anti-OMD 1, Anti-ORM, Anti-Orosomucoid 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

manufacturer/tradename

Genway 15-288-22927F

technique(s)

indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ORM1(5004)

Immunogen

α-1-acid glycoprotein 1 native protein.
Immunogen Sequence: GI # 9257232, sequence 1-201

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crystal D Karakochuk et al.
Diagnostics (Basel, Switzerland), 8(1) (2018-02-03)
Recently, a multiplex ELISA (Quansys Biosciences) was developed that measures ferritin, soluble transferrin receptor (sTfR), retinol-binding protein (RBP), C-reactive protein (CRP), α¹-acid glycoprotein (AGP), thyroglobulin, and histidine-rich protein 2. Our primary aim was to conduct a method-comparison study to compare
Eleanor Brindle et al.
PloS one, 9(12), e115164-e115164 (2014-12-20)
Deficiencies of vitamin A and iron affect a significant portion of the world's population, and efforts to characterize patterns of these deficiencies are hampered by a lack of measurement tools appropriate for large-scale population-based surveys. Vitamin A and iron are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service