Skip to Content
Merck
All Photos(1)

Documents

GW22689B

Sigma-Aldrich

Anti-MMP12 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MMP12(4321)

General description

MMP12 (matrix metalloproteinase 12) is a member of the MMP protein family and is localized to human chromosome 11q22.2-22.3. It is made of nine introns and ten exons, and spans 13kb. It is expressed in a very narrow range of human tissues, and assay of multiple normal human tissues showed its mRNA expression only in placenta. It is found to be expressed in alveolar macrophages of cigarette smokers. It is also called macrophage metalloelastase, and is produced in an inactive form of 54kDa. It is cleaved into an active form of 45kDa, which if further cleaved into 22kDa form. Predominantly, it acts on elastin, and can also degrade fibronectin, laminin, vitronectin etc.

Immunogen

Immunogen Sequence: GI # 4505207, sequence 107-470
Recombinant matrix metalloproteinase 12 preproprotein

Application

Anti-MMP12 antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Matrix metalloproteinase-12 (MMP-12) may act as a specific marker of Stanford-A acute aortic dissection (AAD). The gene is expressed in the primary tumor and acts as a molecular marker that may be useful for predicting extracapsular spread (ECS) in head and neck squamous cell carcinomas (HNSCC) patients with metastatic nodal disease. It may be associated with asthma through inappropriate attraction of leukocytes to the inflamed tissue. It may also serve as a potential diagnostic tool and a therapeutic target in vascular injury and repair.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fumiya Yamaide et al.
International archives of allergy and immunology, 160(3), 287-296 (2012-10-19)
Matrix metalloproteinase 12 gene (MMP12) has been shown to be associated with asthma in a Caucasian population. In this study, we investigate whether single-nucleotide polymorphisms (SNPs) of MMP12 are associated with a risk for asthma in a Japanese population. We
Jin Man Kim et al.
European archives of oto-rhino-laryngology : official journal of the European Federation of Oto-Rhino-Laryngological Societies (EUFOS) : affiliated with the German Society for Oto-Rhino-Laryngology - Head and Neck Surgery, 270(3), 1137-1142 (2012-08-22)
Matrix metalloproteinases (MMPs) play an important role in tumor invasiveness and metastasis. The aim of this study was to investigate the expression pattern of MMPs in the primary tumor of head and neck squamous cell carcinomas (HNSCC) with cervical node
Maria Proietta et al.
Annals of medicine, 46(1), 44-48 (2014-01-18)
The study evaluated macrophage cytokines and macrophage metalloprotease (MMP)-12 levels in patients with Stanford-A acute aortic dissection (AAD) and in patients with critical carotid artery stenosis (CAS) compared with patients matched for age, sex, and traditional cardiovascular risk factors (RF).
A Belaaouaj et al.
The Journal of biological chemistry, 270(24), 14568-14575 (1995-06-16)
Human macrophage metalloelastase (HME) is a recent addition to the matrix metalloproteinase (MMP) family that was initially found to be expressed in alveolar macrophages of cigarette smokers. To understand more about HME expression, analysis of the structure and location of
Yi Song et al.
BMC cardiovascular disorders, 13, 34-34 (2013-05-07)
Aortic dissection(AD) is an acute process of large blood vessels characterized by dangerous pathogenic conditions and high disability and high mortality. The pathogenesis of AD remains debated. Matrix metalloproteinase-12 (MMP-12) participates in many pathological processes such as abdominal aortic aneurysm

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service