Skip to Content
Merck
All Photos(1)

Key Documents

GW22683

Sigma-Aldrich

Anti-ENO1 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$372.00

$372.00


Available to ship on04 April 2025Details


Request a Bulk Order

Select a Size

Change View
50 μG
$372.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$372.00


Available to ship on04 April 2025Details


Request a Bulk Order

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse, rat

manufacturer/tradename

Genway 15-288-22683

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ENO1(2023)

General description

Enolase 1 (ENO1 or ENOα) is a type of enolase enzyme, which in mammals, has three different isoforms namely, α-, β- and γ-enolase. It is an essential cytoplasmic glycolytic enzyme, and has a molecular weight of 48kDa. An alternatively spliced form resides in the nucleus, which has a molecular weight of 37kDa.[1]

Immunogen

Immunogen Sequence: GI # 4503571, sequence 161-320
Recombinant enolase 1

Application

Anti-ENO1 antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Enolase 1(α-enolase) is a homodimer composed of 2 α subunits. ENO1 is involved in tumor progression and prognosis of human glioma. Overexpression of ENO1 is associated with progression of glioma while its knockdown leads to suppressed cell growth, migration and invasion progression by inactivating the PI3K/Akt pathway in glioma cells.[1] The lower expression of ENO1 decreases the response to hypoxia and enhances the sensitivity of the cells to radiation therapy. It may act as a drug target for the treatment of breast cancer.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jie Gao et al.
Oncology reports, 29(4), 1322-1332 (2013-02-06)
In the present study, we investigated the effect of reduced enolase-1 expression in human umbilical vein endothelial cells (HUVECs)/MDA-MB-231 cells on the response to hypoxia and the possible mechanisms involved. Breast cancer cells transfected with enolase-1 siRNA were injected into
Ye Song et al.
Molecular cancer, 13, 65-65 (2014-03-22)
The success of using glycolytic inhibitors for cancer treatment relies on better understanding the roles of each frequently deregulated glycolytic genes in cancer. This report analyzed the involvement of a key glycolytic enzyme, alpha-enolase (ENO1), in tumor progression and prognosis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service