GW22680
Anti-AFP antibody produced in chicken
affinity isolated antibody, buffered aqueous solution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
biological source
chicken
Quality Level
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
species reactivity
human
technique(s)
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... AFP(174)
Application
Anti-AFP antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.
Biochem/physiol Actions
α-Fetoprotein (AFP) is a major serum protein encoded by the AFP gene in humans. It is produced by liver of a fetus. AFP is mapped within bands q11-22 of the long arm of human chromosome 4. Serum α-fetoprotein (AFP) level is elevated in various types of acute liver injuries, indicating active liver regeneration. It is useful for the detection of hepatocellular carcinoma (HCC) recurrence in AFP-producing HCC.
Physical form
Solution in phosphate buffered saline containing 0.02% sodium azide.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
recommended
Product No.
Description
Pricing
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
American journal of human genetics, 35(4), 565-572 (1983-07-01)
Albumin and alpha-fetoprotein are structurally related serum proteins, having a similar gene structure and, conceivably, a common evolutionary origin. To test their relative arrangement in the human genome, the serum albumin and alpha-fetoprotein genes were mapped by in situ hybridization
Journal of gastroenterology and hepatology, 29(1), 157-164 (2013-12-21)
The performance of alpha-fetoprotein (AFP) in the detection of hepatocellular carcinoma (HCC) recurrence after radiofrequency ablation was analyzed. One hundred and forty-six solitary HCC lesions treated by radiofrequency ablation were evaluated. Using the AFP cutoff level at ≥ 20 ng/mL, tumors were
Association between serum alpha-fetoprotein levels and fatty liver disease: a cross-sectional study.
World journal of gastroenterology, 20(33), 11865-11870 (2014-09-11)
To investigate the association between serum alpha-fetoprotein (AFP) levels and fatty liver disease (FLD) in a Chinese population. A cross-sectional study was performed among subjects who presented for a health examination at the First Affiliated Hospital, College of Medicine, Zhejiang
Hepato-gastroenterology, 60(127), 1592-1596 (2014-03-19)
The clinical course of acute viral hepatitis A (AHA) is highly variable. Serum alphafetoprotein (AFP) level is often elevated in various types of acute liver injuries, indicating active liver regeneration. This study was aimed to investigate the clinical significance of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service