Skip to Content
Merck
All Photos(1)

Key Documents

GW22348F

Sigma-Aldrich

Anti-LXN antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$496.00

$496.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 μG
$496.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$496.00


Check Cart for Availability

Request a Bulk Order

biological source

chicken

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse, rat

manufacturer/tradename

Genway 15-288-22348F

technique(s)

indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LXN(56925)

General description

LXN (latexin) is the human homologue of endogenous inhibitor of carboxypeptidase A found in rat. It was initially recognized as a neural marker in developing mammalian brain, of the lateral neocortex.[1] This protein is composed of 222 amino acids. An α-helix links two subdomains that comprise the protein. It has high structural homology to TIG1 (tazarotene-induced gene 1).

Immunogen

Immunogen Sequence: GI # 9910396, sequence 1-222
Recombinant latexin protein

Application

Anti-LXN antibody produced in chicken is suitable for indirect ELISA and western blotting.

Biochem/physiol Actions

Latexin (LXN) is a negative regulator of hematopoietic stem cell and may exert its tumor-suppressive function by altering the stem cell-like properties of melanoma cells.[1] Its expression is reduced in human gastric cancers compared with their normal control tissues. LXN expression may have involvement in tumor suppression. It may have clinical efficacy in the treatment of malignancies.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi Liu et al.
PloS one, 7(9), e44979-e44979 (2012-10-03)
Latexin is a negative regulator of hematopoietic stem cell number in mice. Its dysregulated expression in other tumors led us to hypothesize that latexin may have tumor suppressor properties in hematological malignancies. We found that latexin was down-regulated in a
Viswanathan Muthusamy et al.
The Journal of investigative dermatology, 133(7), 1827-1833 (2013-02-01)
Despite recent advancements in therapy, melanoma remains a highly lethal skin cancer. A better understanding of the genetic and epigenetic changes responsible for melanoma formation and progression could result in the development of more effective treatments. Advanced melanomas are known
Yong Li et al.
BMC cancer, 11, 121-121 (2011-04-07)
Latexin, also known as endogenous carboxypeptidase inhibitor (CPI), has been found to inhibit mouse stem cell populations and lymphoma cell proliferation, demonstrating its potential role as a tumor suppressor. Our previous study also suggested a correlation between latexin expression and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service