Skip to Content
Merck
All Photos(1)

Key Documents

GW22280A

Sigma-Aldrich

Anti-KLHL12 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-C3IP1, Anti-DKIR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

manufacturer/tradename

Genway 15-288-22280A

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KLHL12(59349)

General description

Kelch-like protein 12 is a novel human kelch protein localized to a ring canal structure. It consists of N-terminal POZ (poxviruses and zinc fingers) and intervening region (IVR) domains. It plays an important role in the ring-like structure activity during oocyte development.

Immunogen

Immunogen Sequence: GI # 11056006, sequence 205-404
Recombinant kelch-like 12

Application

Anti-KLHL12 antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Kelch-like protein 12 is a protein encoded by the KLHL12 gene in humans. It is a Bric à Brac (BTB)-Kelch protein and specifically binds to the polymorphic region of the D4 receptor through its Kelch domain. KLHL12 also interacts with Cullin3 and functions as an adaptor to target the D4 receptor to an E3 ubiquitin ligase complex. Its overexpression strongly promotes ubiquitination of the D4 receptor. KLHL12 is essential for the development of the ring-like structure, which is independent of the ability to bind actin. It interacts with both immature, ER-associated and mature, plasma membrane-associated D4 receptors, β-arrestin2, and also promotes ubiquitination of both receptor subpools.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angela Mai et al.
Experimental cell research, 300(1), 72-83 (2004-09-24)
We have previously purified and cloned an apoptosis-inducing protein (AIP) derived from fish infected with the anisakis simplex. Recently, we identified a series of AIP-responsive genes in the HL-60 cell line using a subtractive hybridization method. Here we report the
Pieter Rondou et al.
Cellular signalling, 22(6), 900-913 (2010-01-27)
In previous studies, we identified KLHL12 as a novel interaction partner of the dopamine D4 receptor that functions as an adaptor in a Cullin3-based E3 ubiquitin ligase complex to target the receptor for ubiquitination. In this study, we show that
Pieter Rondou et al.
The Journal of biological chemistry, 283(17), 11083-11096 (2008-02-28)
Dopamine receptors belong to the superfamily of G-protein-coupled receptors and are subdivided into D1-type (D1 and D5) and D2-type (D2, D3, and D4) receptors. The D4 receptor has a remarkable polymorphism in its third intracellular loop, which is under intensive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service