Skip to Content
Merck
All Photos(1)

Key Documents

GW22261

Sigma-Aldrich

Anti-HADHSC antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$422.00

$422.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 μG
$422.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$422.00


Check Cart for Availability

Request a Bulk Order

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, mouse, human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HADH(3033)

General description

Chicken polyclonal anti-HADHSC antibody reacts with mouse, rat, human L-3-hydroxyacyl-Coenzyme A dehydrogenase, short chain, EC 1.1.1.35, enzymes.
L-3-hydroxyacyl-Coenzyme A dehydrogenase, short chain (EC 1.1.1.35) is an enzyme that plays an essential role in the mitochondrial β-oxidation of short chain fatty acids. It exerts it highest activity toward 3-hydroxybutyryl-CoA.

Immunogen

Immunogen Sequence: GI # 4885387, sequence 57-314
Recombinant L-3-hydroxyacyl-Coenzyme A dehydrogenase, short chain

Application

Chicken polyclonal anti-HADHSC antibody is used to tag medium and short chain L-3-hydroxyacyl-coenzyme A dehydrogenase, EC 1.1.1.35, for detection and quantitation by immunocytochemical and immunohistochemical (IHC) techniques. It is used as a probe to determine the presence and roles of medium and short chain L-3-hydroxyacyl-coenzyme A dehydrogenase in mitochondrial β-oxidation of short chain fatty acids.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service