Skip to Content
Merck
All Photos(1)

Key Documents

GW21469

Sigma-Aldrich

Anti-PTPRF antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Receptor-type tyrosine-protein phosphatase F

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$372.00

$372.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 μG
$372.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$372.00


Check Cart for Availability

Request a Bulk Order

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, human, mouse

manufacturer/tradename

Genway 15-288-21469

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PTPRF(5792)

Immunogen

Immunogen Sequence: GI # 4506311, sequence 1151-1225
Recombinant protein tyrosine phosphatase, receptor type, F isoform 1

Application

Anti-PTPRF antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Receptor-type tyrosine-protein phosphatase F is an enzyme encoded by the PTPRF gene in humans. It localizes to adherens junctions and may be involved in the regulation of epithelial cell-cell contacts, peptide growth factor signaling and the canonical Wnt pathway. PTPRF induction occurs by cell-cell contact during cell proliferation that activates ERK-dependent proliferation signaling to prevent cell hyperproliferation and tumor initiation. It acts as a crucial regulator of the sensitivity of two key insulin signalling pathways to insulin. PTPRF regulates cell proliferation, response to growth factors and cellular adhesion. Mutation in the gene is associated with breast cancer.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guntram Borck et al.
Human genetics, 133(8), 1041-1047 (2014-05-02)
Athelia is a very rare entity that is defined by the absence of the nipple-areola complex. It can affect either sex and is mostly part of syndromes including other congenital or ectodermal anomalies, such as limb-mammary syndrome, scalp-ear-nipple syndrome, or
Rabindranath Bera et al.
Hepatology (Baltimore, Md.), 59(6), 2238-2250 (2014-01-29)
It is unclear how proliferating cells elicit suppression on cell proliferation and how cancer cells evade this growth suppression. Using a loss-of-function screening of the human kinome and phosphatome to identify genes suppressing tumor initiation in human hepatocellular carcinoma (HCC)
T Yang et al.
Molecular carcinogenesis, 25(2), 139-149 (1999-06-12)
The findings that protein tyrosine phosphatases (PTPs) regulate cell proliferation, response to growth factors, and cellular adhesion and the discovery that mutations in PTP genes are associated with breast cancer suggest that altered expression of PTPs contributes to the breast
Ann Mander et al.
FEBS letters, 579(14), 3024-3028 (2005-05-18)
To test the role of the leukocyte common antigen-related protein tyrosine phosphatase (LAR) as a regulator of insulin receptor (IR) signalling, an siRNA probe against LAR was developed. Knock-down of LAR induced post-receptor insulin resistance with the insulin-induced activation of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service