Skip to Content
Merck
All Photos(1)

Key Documents

GW20083F

Sigma-Aldrich

Anti-Human IgG (Fc specific) antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Immunoglobulin G Fc

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$518.00

$518.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 μG
$518.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

$518.00


Check Cart for Availability

Request a Bulk Order

biological source

chicken

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, human, mouse

manufacturer/tradename

Genway 15-288-20083F

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IGHG1(3500)

Immunogen

IgG Fc human native protein.

Application

Anti-Human IgG (Fc specific) antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Ig γ-1 chain C region is a protein encoded by the IGHG1 gene in humans. Igγ-1 chain C region is present in human pancreatic cancer tissues and its expression is very low in adjacent noncancerous tissues. IGHG1 blockage leads to retarded tumor growth and improved survival. Its presence in pancreatic cancer cells might constitute an important element that is responsible for tumor cell proliferation and immune evasion mechanisms. IGHG1 is associated with prostate cancer and can inhibit cell viability leading to apoptosis. It may act as a potential target in prostate cancer gene therapy.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Judith Blaine
Methods in molecular biology (Clifton, N.J.), 2224, 123-132 (2021-02-20)
Proteinuria is a widely used marker of renal disease and is strongly associated with renal and cardiovascular outcomes. The molecular mechanisms underlying filtration of serum proteins through the glomerular filtration barrier (GFB) remain to be determined. Since the GFB is
Bin Pan et al.
Molecular biology reports, 40(1), 27-33 (2012-11-03)
To investigate the immunoglobulin G (IgG) expression in prostate cancer cell lines and explore the effects of IGHG1 gene knockdown on PC3 cell growth and apoptosis. Flow cytometry, qPCR and western blot were used to demonstrate IgG expression in prostate
Xiaoyan Li et al.
Pancreas, 40(5), 753-761 (2011-06-10)
To investigate the expression of Igγ-1 chain C region (IGHG1) in human pancreatic carcinomas and determine the biological function of IGHG1 expression in immune evasion mechanisms. Comparative proteomic analysis was used to detect the differential expression of IGHG1 in human
James Dylewski et al.
PloS one, 14(2), e0209732-e0209732 (2019-02-28)
Proteinuria is strongly associated with kidney disease progression but the mechanisms underlying podocyte handling of serum proteins such as albumin and IgG remain to be elucidated. We have previously shown that albumin and IgG are transcytosed by podocytes in vitro.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service