Skip to Content
Merck
All Photos(1)

Key Documents

GW20025F

Sigma-Aldrich

Anti-GM-CSF antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CSF, Anti-Colony-stimulating factor, Anti-Granulocyte-Macrophage Colony Stimulating Factor, Anti-MGC131935

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$389.00

$389.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
50 μG
$389.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$389.00


Estimated to ship on25 May 2025


Request a Bulk Order

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, human, mouse

manufacturer/tradename

Genway 15-288-20025F

technique(s)

indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CSF2(1437)

General description

Granulocyte-macrophage colony-stimulating factor (GM-CSF), also known as CSF-2, is encoded by the gene mapped to human chromosome 5q31-33. The encoded protein is expressed in a wide variety of immune cell types, including macrophages, mast cells and T cells, mainly in response to immune activation and inflammatory cytokines. In addition, it is also secreted in various non-immune cell types, such as endothelial cells, keratinocytes and fibroblasts, following the appropriate stimuli.

Immunogen

Immunogen Sequence: GI # 27437030, sequence 1-144
Recombinant colony stimulating factor 2

Application

Anti-GM-CSF antibody produced in chicken is suitable for western blotting at a working dilution of 1:500 and for cell staining at a working dilution of 1:200. It is also suitable for indirect ELISA.

Biochem/physiol Actions

GM-CSF (Granulocyte-macrophage colony-stimulating factor) is a pleiotropic cytokine that regulates the growth and differentiation of granulocytes, macrophages, eosinophils and erythrocytes. The gene is present in a cluster along with interleukin-3 (IL-3), IL-4 and IL-5 on chromosome 5.
Granulocyte-macrophage colony-stimulating factor (GM-CSF) facilitates binding of eosinophils to vascular cell adhesion molecule-1 and eosinophil migration into tissues. In addition, it is also implicated in maturation of antigen presenting cells. GM-CSF plays an essential role in modulation of surfactant homeostasis and alveolar macrophage innate immune responses in the lung. It also plays a vital role immune-independent tumor progression. Mutation in the gene is associated with the development of atopic asthma.
Granulocyte-macrophage colony-stimulating factor (GM-CSF) is used in the treatment of neutropenia and leukemia in combination with chemotherapies. It plays a major role in proliferation and differentiation of granulocytes and macrophages from bone marrow progenitor cells. GM-CSF also play a vital role in immune regulation and hematopoiesis.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B H van Leeuwen et al.
Blood, 73(5), 1142-1148 (1989-04-01)
The human genes for the hematopoietic growth factors interleukin-3 (IL-3), IL-5, and granulocyte-macrophage colony-stimulating factor (GM-CSF) have been mapped to 5q23-31. We present in situ hybridization evidence that the human IL-4 gene is located at 5q23.3-31.2, suggesting that the four
K Kaushansky et al.
Proceedings of the National Academy of Sciences of the United States of America, 83(10), 3101-3105 (1986-05-01)
Through the use of long single-sequence oligonucleotide probes, the complete gene for human granulocyte-macrophage colony-stimulating factor (hGM-CSF) has been cloned from a human genomic library. The gene is 2.5 kilobases in length, contains three introns, and is present as a
Sabine Hoffjan et al.
American journal of human genetics, 76(4), 696-704 (2005-02-24)
Asthma is a common disease that results from both genetic and environmental risk factors. Children attending day care in the 1st year of life have lower risks for developing asthma, although the mechanism for this "day care" effect is largely

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service