Key Documents
GN701061
Greiner Multiwell Strips
high binding, wells flat bottom (1 × 12 well)
Synonym(s):
Greiner Multiwell Strips
About This Item
UNSPSC Code:
41121800
Pricing and availability is not currently available.
packaging
pack of 400 ea
manufacturer/tradename
Greiner 701061
availability
available only in China
General description
Both medium-binding and high binding are offered. High binding plates and strips are recommended for ELISAs. High binding denotes a binding capacity of 600 ng/cm2 and scatter of less than 10%. Optimal protein binding is variable and comparative tests between high and medium binding are suggested.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)