Skip to Content
Merck
All Photos(1)

Key Documents

GERPN810

ECL DualVue Western Markers

Cytiva RPN810, sufficient for 25 blots

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$294.00

$294.00


Check Cart for Availability


Select a Size

Change View
1 PKG
$294.00

About This Item

UNSPSC Code:
41105331
NACRES:
NA.25

$294.00


Check Cart for Availability

form

solution

usage

sufficient for 25 blots

feature

wetted part: no

manufacturer/tradename

Cytiva RPN810

packaging

pkg of 25 loadings

shipped in

dry ice

storage temp.

−20°C

Related Categories

General description

ECL DualVue Western Blotting Markers contain both colored proteins and tagged proteins that introduce convenience and reliability and facilitate accurate size determination of proteins on Western blots.

Application

Amersham ECL DualVue Western Blotting Markers perform two key functions. First, the prestained marker Proteins confirm Protein transfer in addition to clearly defining blot orientation. Second, the recombinant tagged Proteins are detected on film or by CCD imaging in conjunction with the target Protein(s) of interest, enabling highly accurate molecular mass determination of the target protein(s). The tagged markers are detected easily and specifically by means of a specific conjugate that eliminates any cross contamination between target Protein(s) and markers.

Amersham ECL DualVue Western Blotting Markers contain a mixture of three prestained colored markers of defined molecular weight (from 15 000 to 100 000) and seven tagged recombinant Proteins (molecular weights from 15 000 to 150 000). Each pack contains ECL DualVue markers (125 μL) and S-Protein-HRP conjugate (50 μL), sufficient for 25 standard gel loadings.

Features and Benefits

  • Detect Proteins on gel and membrane as well as on film.
  • Prestained markers indicate efficiency of Protein transfer to membrane and blot orientation.
  • Tagged recombinant Protein markers allow accurate calculation of molecular weight of target Protein(s).
  • Product is compatible with any HRP substrate as well as both PVDF and nitrocellulose membrane types.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Amersham is a trademark of Cytiva
ECL is a trademark of Cytiva

Storage Class Code

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

John Kerwin et al.
eLife, 9 (2020-04-22)
As part of the Reproducibility Project: Cancer Biology we published a Registered Report (Khan et al., 2015), that described how we intended to replicate selected experiments from the paper "A coding-independent function of gene and pseudogene mRNAs regulates tumour biology"

Articles

This page describes troubleshooting strategies in the detection of GST-tagged proteins.

Protocols

This page describes chemiluminescence detection of GST-tagged proteins with Amersham ECL, Amersham ECL Prime, and Amersham ECL Select from Cytiva.

This page describes how to monitor expression and purification of GST-tagged proteins by Western blot analysis using Cytiva products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service