Skip to Content
Merck
All Photos(1)

Key Documents

GEE70092X

Shrimp Alkaline Phosphatase

Cytiva E70092X, pack of 5000 U

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$1,080.00

$1,080.00


Check Cart for Availability


Select a Size

Change View
1 PKG
$1,080.00

About This Item

UNSPSC Code:
12352204
NACRES:
NA.54

$1,080.00


Check Cart for Availability

packaging

pack of 5000 U

manufacturer/tradename

Cytiva E70092X

shipped in

dry ice

storage temp.

−20°C

Related Categories

General description

A high specific activity, heat-labile alkaline phosphatase. Completely and irreversibly inactivated in Tris buffers at pH 8.0 to 8.5 by heating for 15 min at 65°C. No further treatment is necessary.

Application

Shrimp alkaline phosphatase has been used to degrade primers from amplified samples after PCR.[1][2]

Features and Benefits

  • Removing 5′-phosphates from DNA and RNA.
  • Easily inactivated by heat.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Storage Class Code

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mitochondrial DNA variation in the eastern pondmussel, Ligumia nasuta (Bivalvia: Unionoida), in the Great Lakes Region.
Scott M W, et al.
Walkerana, 17(2), 60-60 (2014)
Mitochondrial DNA structure of Pyganodon grandis (Bivalvia: Unionidae) from the Lake Erie watershed and selected locations in its northern distribution.
Krebs R A, et al.
American Malacological Union, 33(1), 34-42 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service